Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01590063_s1
          See other LOC100289094 GE Assays ›
          Gene Symbol:
          LOC100289094
          Gene Name:
          uncharacterized LOC100289094
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01590063_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100289094
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC025775.1 - 1 722 107

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 605731 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 121055 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P491499 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P491499 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 15019 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 12000 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P491499 - -
          Agilent Human 1B Microarray G4111A A_32_P491499 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011371.hg.1 ENST00000600041.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000407635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200011371.hg.1 ENST00000600041.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1559877_PM_at - -
          Human Genome U133 Plus 2.0 Array 1559877_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559877_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559877_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559877_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559877_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559877_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559877_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559877_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559877_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559877_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559877_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559877_at9 - -
          Human Genome U95 Set 66439_at - -
          Human Genome U95 Set 66439_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229362 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1829502 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 870274 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_75789 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1818213 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1818213 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1829502 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline