| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006624.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006624.hg.1 |
TC09002907.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006630.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006630.hg.1 |
ENST00000538816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006630.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006630.hg.1 |
TC09002907.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006628.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006628.hg.1 |
ENST00000538816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006628.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006628.hg.1 |
TC09002907.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006631.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006631.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006631.hg.1 |
ENST00000538816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006631.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006631.hg.1 |
TC09002907.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006632.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006632.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006632.hg.1 |
ENST00000538816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006632.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006632.hg.1 |
TC09002907.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006625.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006625.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006625.hg.1 |
TC09002907.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184476.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184476.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184476.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184476.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184476.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184476.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184476.hg.1 |
TC0900012162.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012162.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184471.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184471.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184471.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184471.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184471.hg.1 |
TC0900012162.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900184473.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184473.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184473.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184473.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184473.hg.1 |
TC0900012162.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900184470.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184470.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184470.hg.1 |
TC0900012162.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900184474.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184474.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184474.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184474.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184474.hg.1 |
TC0900012162.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900184475.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184475.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184475.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184475.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184475.hg.1 |
TC0900012162.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
ENST00000431067.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012163.hg.1 |
NM_000608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184477.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184477.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184477.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184477.hg.1 |
ENST00000259396.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184477.hg.1 |
ENST00000477456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184477.hg.1 |
NM_000607.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184477.hg.1 |
TC0900012162.hg.1 |
Clariom D TC |