Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01592552_m1
          See other METTL21A GE Assays ›
          Gene Symbol:
          METTL21A
          Gene Name:
          methyltransferase 21A, HSPA lysine
          Gene Aliases:
          FAM119A, HCA557b, HSPA-KMT
          Chromosome Location:
          Chr.2: 207580635 - 207626053 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01592552_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: METTL21A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127395.5 NP_001120867.1 2 - 3 516 125
          NM_001308021.3 NP_001294950.1 2 - 3 516 125
          NM_001330130.2 NP_001317059.1 2 - 3 313 125
          NM_001330131.2 NP_001317060.1 2 - 3 516 125
          NM_001330132.3 NP_001317061.1 2 - 3 444 125
          NM_001330133.3 NP_001317062.1 2 - 3 444 125
          NM_001330134.2 NP_001317063.1 2 - 3 560 125
          NM_001330135.2 NP_001317064.1 2 - 3 313 125
          NM_001330136.3 NP_001317065.1 2 - 3 323 125
          NM_001330137.2 NP_001317066.1 2 - 3 330 125
          NM_001388427.1 NP_001375356.1 2 - 3 516 125
          NM_001388439.2 NP_001375368.1 1 - 2 397 125
          NM_001393563.1 NP_001380492.1 1 - 2 397 125
          NM_001393564.1 NP_001380493.1 2 - 3 274 125
          NM_001393565.1 NP_001380494.1 1 - 2 397 125
          NM_001393566.1 NP_001380495.1 2 - 3 313 125
          NM_001393567.1 NP_001380496.1 2 - 3 313 125
          NM_001393568.1 NP_001380497.1 1 - 2 397 125
          NM_001393569.1 NP_001380498.1 2 - 3 516 125
          NM_001393570.1 NP_001380499.1 2 - 3 313 125
          NM_001393571.1 NP_001380500.1 2 - 3 516 125
          NM_001393572.1 NP_001380501.1 2 - 3 313 125
          NM_001393573.1 NP_001380502.1 2 - 3 274 125
          NM_001393574.1 NP_001380503.1 1 - 2 397 125
          NM_001393575.1 NP_001380504.1 2 - 3 323 125
          NM_145280.7 NP_660323.3 2 - 3 330 125
          XM_005246336.4 XP_005246393.1 2 - 3 516 125
          XM_005246337.5 XP_005246394.1 2 - 3 313 125
          XM_006712327.4 XP_006712390.1 2 - 3 560 125
          XM_011510724.3 XP_011509026.1 2 - 3 274 125
          XM_011510725.3 XP_011509027.1 2 - 3 274 125
          XM_011510728.4 XP_011509030.1 2 - 3 274 125
          XM_024452721.2 XP_024308489.1 2 - 3 330 125
          XM_024452722.2 XP_024308490.1 2 - 3 313 125
          XM_047443496.1 XP_047299452.1 2 - 3 274 125
          XM_047443497.1 XP_047299453.1 2 - 3 560 125
          XM_047443498.1 XP_047299454.1 2 - 3 274 125
          XM_047443499.1 XP_047299455.1 2 - 3 313 125
          GenBank mRNA AF455817.1 - 2 - 3 265 125
          AK093812.1 - 2 - 3 314 125
          BC009462.1 - 2 - 3 453 125
          BC033720.1 - 2 - 3 325 125
          BG832122.1 - 2 - 3 371 125
          CN367990.1 - 2 - 3 252 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200015548.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002712.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene METTL21A

          Gene Ontology Categories:

          Function(s) Process(es)

          protein methylation
          methylation
          negative regulation of protein folding
          protein binding
          methyltransferase activity
          protein methyltransferase activity
          protein-lysine N-methyltransferase activity
          transferase activity
          Hsp70 protein binding
          heat shock protein binding
          ATPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000272839.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000272839.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000272839.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000272839.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000272839.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000272839.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000272839.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000272839.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000406927.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000406927.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000406927.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000406927.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000406927.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000406927.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000406927.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000406927.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000411432.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000425132.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000425132.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000425132.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000425132.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000425132.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000425132.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000425132.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000426075.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000426075.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000426075.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000426075.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000426075.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000426075.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000432416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000432416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000432416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000432416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000432416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000442521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000442521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000442521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000442521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000442521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448007.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448007.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448007.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448007.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448007.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448007.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448007.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448007.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000448823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000458426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000458426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000458426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000458426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000458426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000458426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000458426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000458426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000458426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000461419.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000461419.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000461419.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000471128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000471128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000471128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000471128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000471128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000477919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000477919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000477919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000477919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000477919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 ENST00000477919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001127395.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001308021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001308021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001308021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001308021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001308021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001308021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_001308021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_145280.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_145280.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_145280.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_145280.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_145280.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_145280.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_145280.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_145280.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_145280.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_145280.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015548.hg.1 NM_145280.6 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline