Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01594707_m1
          See other PIP4K2B GE Assays ›
          Gene Symbol:
          PIP4K2B
          Gene Name:
          phosphatidylinositol-5-phosphate 4-kinase type 2 beta
          Gene Aliases:
          PI5P4KB, PIP5K2B, PIP5KIIB, PIP5KIIbeta, PIP5P4KB
          Chromosome Location:
          Chr.17: 2801010 - 2834875 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01594707_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PIP4K2B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003559.5 NP_003550.1 5 - 6 782 148
          XM_011525326.4 XP_011523628.1 6 - 7 863 148
          XM_011525327.3 XP_011523629.1 7 - 8 994 148
          XM_011525330.2 XP_011523632.1 4 - 5 467 148
          XM_047436888.1 XP_047292844.1 7 - 8 1023 148
          XM_047436889.1 XP_047292845.1 6 - 7 942 148
          XM_047436890.1 XP_047292846.1 3 - 4 391 148
          GenBank mRNA BC027459.1 - 5 - 6 748 148
          BT019614.1 - 5 - 6 650 148
          U85245.1 - 5 - 6 1130 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010545.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001433.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PIP4K2B

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          cell surface receptor signaling pathway
          regulation of autophagy
          phosphatidylinositol metabolic process
          negative regulation of insulin receptor signaling pathway
          phosphatidylinositol phosphorylation
          autophagosome-lysosome fusion
          1-phosphatidyl-1D-myo-inositol 4,5-bisphosphate biosynthetic process
          positive regulation of autophagosome assembly
          nucleotide binding
          protein binding
          ATP binding
          GTP binding
          kinase activity
          1-phosphatidylinositol-4-phosphate 5-kinase activity
          1-phosphatidylinositol-5-phosphate 4-kinase activity
          transferase activity
          protein homodimerization activity
          phosphatidylinositol kinase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17018928.hg.1 ENST00000269554.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17018928.hg.1 ENST00000311500.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17018928.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17018928.hg.1 TC17001433.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010545.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700182997.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700182997.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700182997.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700182997.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700182997.hg.1 ENST00000613767.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700182997.hg.1 ENST00000617781.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700182997.hg.1 ENST00000619039.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700182997.hg.1 NM_003559.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700182997.hg.1 TC1700010545.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0