Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01596743_m1
          See other POMC GE Assays ›
          Gene Symbol:
          POMC
          Gene Name:
          proopiomelanocortin
          Gene Aliases:
          ACTH, CLIP, LPH, MSH, NPP, POC
          Chromosome Location:
          Chr.2: 25160853 - 25168851 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01596743_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: POMC
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000939.3 NP_000930.1 2 - 3 508 142
          NM_001035256.2 NP_001030333.1 3 - 4 558 142
          NM_001319204.1 NP_001306133.1 3 - 4 588 142
          NM_001319205.1 NP_001306134.1 2 - 3 538 142
          GenBank mRNA BC065832.1 - 2 - 3 220 30392189 142
          CD106051.1 - 3 - 4 324 142
          CD106168.1 - 280 142
          CD108235.1 - 2 - 3 231 142
          CR541826.1 - 1 - 2 134 142
          DQ884404.1 - 1 - 2 134 142
          M38297.1 - 1 - 2 119 142
          S76492.1 - 1 - 2 38 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200011998.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001657.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the S76492.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene POMC

          Gene Ontology Categories:

          Function(s) Process(es)

          generation of precursor metabolites and energy
          signal transduction
          neuropeptide signaling pathway
          cell-cell signaling
          regulation of blood pressure
          peptide hormone processing
          regulation of appetite
          negative regulation of tumor necrosis factor production
          cellular pigmentation
          glucose homeostasis
          positive regulation of transcription from RNA polymerase II promoter
          regulation of glycogen metabolic process
          regulation of corticosterone secretion
          G-protein coupled receptor binding
          receptor binding
          hormone activity
          neuropeptide hormone activity
          type 3 melanocortin receptor binding
          type 4 melanocortin receptor binding
          type 1 melanocortin receptor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000380794.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000395826.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000395826.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000395826.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000395826.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000395826.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000395826.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000395826.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000395826.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000395826.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000395826.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000395826.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000449220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000449220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000449220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000449220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000449220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000449220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000449220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 ENST00000449220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_000939.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011998.hg.1 NM_001035256.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200187247.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200187247.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200187247.hg.1 ENST00000264708.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200187247.hg.1 ENST00000405623.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200187247.hg.1 TC0200011998.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 149988 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline