Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01596838_g1
          See other RBPMS2 GE Assays ›
          Gene Symbol:
          RBPMS2
          Gene Name:
          RNA binding protein, mRNA processing factor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 64739891 - 64775589 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01596838_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RBPMS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_194272.3 NP_919248.1 4 - 5 532 98
          NR_138350.2 4 - 5 532 98
          NR_138363.2 4 - 5 263 98
          NR_138364.2 4 - 5 262 98
          XM_047432446.1 XP_047288402.1 4 - 5 193 98
          GenBank mRNA AK124123.1 - 4 - 5 262 98
          AK127873.1 - 4 - 5 530 98
          AY369207.1 - 4 - 5 514 98
          BC131612.1 - 3 - 4 166 98
          BC148208.1 - 3 - 4 166 98
          DA564899.1 - 4 - 5 256 98
          DA565836.1 - 4 - 5 295 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500009756.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001553.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC131612.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RBPMS2

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          negative regulation of BMP signaling pathway
          embryonic digestive tract morphogenesis
          positive regulation of smooth muscle cell proliferation
          negative regulation of smooth muscle cell differentiation
          nucleic acid binding
          RNA binding
          mRNA binding
          protein binding
          identical protein binding
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15014764.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15014764.hg.1 ENST00000560606.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15014764.hg.1 NM_194272.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15014764.hg.1 TC15001553.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500172071.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1500172071.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500172071.hg.1 NM_194272.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500172071.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1500172071.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500172071.hg.1 NM_194272.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500172071.hg.1 TC1500009756.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline