Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01597103_g1
          See other OR4C13 GE Assays ›
          Gene Symbol:
          OR4C13
          Gene Name:
          olfactory receptor family 4 subfamily C member 13
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 49952391 - 49953419 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01597103_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR4C13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001955.2 NP_001001955.2 1 676 121
          GenBank mRNA BC137005.1 - 1 676 9020998 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100007554.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000422.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR4C13

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07804958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28211999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07804953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16449753 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212028 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3331761 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0202 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0201 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0200 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43060 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P32907034 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3331761 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3331761 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28211999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804954 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212008 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212010 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804954 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212008 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28211999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212010 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07804954 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07804951 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07804952 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07804956 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07804950 - -
          Agilent Human 1B Microarray G4111A A_32_P496496 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16449750 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16449793 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07804951 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07804958 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07804956 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001001955_at - -
          Affymetrix Human Gene 1.0 ST Array 7948047 - -
          Affymetrix Human Gene 1.0 ST Array 7939901 - -
          Affymetrix Human Gene 1.1 ST Array 7948046 - -
          Affymetrix Human Gene 1.1 ST Array 7939900 - -
          Affymetrix Human Genome U219 Array 11742361_at - -
          Affymetrix Human Genome U219 Array 11742361_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11005451.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005451.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005451.hg.1 TC11000422.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100007554.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007554.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007554.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007554.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007554.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007554.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010819.hg.1 ENST00000533375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010780.hg.1 ENST00000532368.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007554.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007554.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007554.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007554.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Clariom D Assay TC1100007554.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Clariom D Assay TC1100007554.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010780.hg.1 ENST00000532368.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153653.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153653.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100153653.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153653.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100153653.hg.1 TC1100007554.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100010819.hg.1 ENST00000533375.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153652.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153652.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100153652.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153652.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100153652.hg.1 TC1100007554.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100153651.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153651.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100153651.hg.1 ENST00000555099.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153651.hg.1 NM_001001955.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100153651.hg.1 TC1100007554.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046368 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046368 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 216281 - -
          ABI Human Genome Survey Microarray Version 2.0 665879 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1730984 NM_001001955.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1730984 BC137005.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1730984 NM_001001955.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1730984 BC137005.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1730984 NM_001001955.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1730984 BC137005.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1730984 NM_001001955.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1730984 BC137005.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1730984 NM_001001955.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1730984 BC137005.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_16650-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1730984 NM_001001955.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1730984 BC137005.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1730984 NM_001001955.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1730984 BC137005.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_16650 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_16650 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1730984 NM_001001955.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1730984 BC137005.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1730984 NM_001001955.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1730984 BC137005.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1730984 NM_001001955.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1730984 BC137005.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1730984 NM_001001955.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1730984 BC137005.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1730984 NM_001001955.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1730984 BC137005.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline