Search
Search

KRTAP10-1| Gene Symbol: KRTAP10-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198691.2 | NP_941964.2 | 1 | 956 | 94 | |
| GenBank mRNA | AB076347.1 | - | 1 | 956 | 94 | |
| AJ566380.1 | - | 71 | 94 | |||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075023 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412720 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412729 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075034 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412731 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17242106 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412725 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412722 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11412726 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075017 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075031 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32075035 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17242104 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3293798 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3374 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 57160 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3293798 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3293798 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075028 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242093 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412729 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242107 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242106 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242108 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075034 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412732 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075016 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242104 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412728 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412721 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412733 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075017 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412727 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075027 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075031 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075023 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412731 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412722 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412720 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242113 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075033 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412734 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412726 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412725 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412735 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075035 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17242108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075017 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075035 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17242104 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075031 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17242093 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17242106 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412731 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412722 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075023 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412720 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412727 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412721 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075017 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412735 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242093 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075016 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075028 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412733 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075035 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242107 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242108 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075027 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075031 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412725 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075033 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242104 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242106 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412728 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412732 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412729 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412734 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17242113 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P32075034 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P11412726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17242104 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075031 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17242093 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17242108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075017 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075035 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17242106 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32075034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11412728 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412735 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412726 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412728 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412731 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412725 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412721 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17242108 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412734 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412729 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412722 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11412720 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_198691_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8070776 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8070775 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8070774 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8070773 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8070772 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8070771 | - | - |
| Affymetrix Human Genome U219 Array | 11715174_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11715174_s_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006297.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006297.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006297.hg.1 | TC21000527.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006295.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006295.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006295.hg.1 | TC21000527.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006296.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006296.hg.1 | ENST00000545982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006296.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006296.hg.1 | TC21000527.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008375.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008375.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008375.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008375.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008375.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008375.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008375.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008375.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008375.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156742.hg.1 | TC2100008375.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100156742.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156742.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156742.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156742.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156741.hg.1 | TC2100008375.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100156741.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156741.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156741.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156741.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008375.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008375.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008375.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008375.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008375.hg.1 | ENST00000400375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008375.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008375.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008375.hg.1 | NM_198691.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008375.hg.1 | NM_198691.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A018975 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A018975 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1771174 | AB076347.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1771174 | BC120959.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1771174 | BC120960.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1771174 | AB076347.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1771174 | BC120959.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1771174 | BC120960.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1771174 | AB076347.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1771174 | BC120959.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1771174 | BC120960.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1771174 | AB076347.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1771174 | BC120959.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1771174 | BC120960.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1771174 | AB076347.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1771174 | BC120959.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1771174 | BC120960.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_18984-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1771174 | AB076347.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1771174 | BC120959.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1771174 | BC120960.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1771174 | AB076347.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1771174 | BC120959.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1771174 | BC120960.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_18984 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_18984 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1771174 | AB076347.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1771174 | BC120959.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1771174 | BC120960.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1771174 | AB076347.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1771174 | BC120959.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1771174 | BC120960.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1771174 | AB076347.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1771174 | BC120959.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1771174 | BC120960.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1771174 | AB076347.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1771174 | BC120959.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1771174 | BC120960.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1771174 | NM_198691.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1771174 | AB076347.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1771174 | AJ566380.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1771174 | BC120959.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1771174 | BC120960.1 | human_transcriptome |