Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01651252_s1
          See other KRTAP12-4 GE Assays ›
          Gene Symbol:
          KRTAP12-4
          Gene Name:
          keratin associated protein 12-4
          Gene Aliases:
          KRTAP12.4
          Chromosome Location:
          Chr.21: 44654213 - 44654659 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01651252_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KRTAP12-4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198698.1 NP_941971.1 1 388 86
          GenBank mRNA AB076360.1 - 1 388 86
          BC125198.1 - 1 388 40123223 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100008379.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000532.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KRTAP12-4

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11413041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11413044 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11413042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11413043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17242360 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17242359 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32075505 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3354394 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3378 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3379 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2575 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3354394 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3354394 - -
          Agilent Whole Chr array_18 19 21 A_17_P17242357 - -
          Agilent Whole Chr array_18 19 21 A_17_P17242360 - -
          Agilent Whole Chr array_18 19 21 A_17_P32075519 - -
          Agilent Whole Chr array_18 19 21 A_17_P11413043 - -
          Agilent Whole Chr array_18 19 21 A_17_P11413044 - -
          Agilent Whole Chr array_18 19 21 A_17_P32075505 - -
          Agilent Whole Chr array_18 19 21 A_17_P11413045 - -
          Agilent Whole Chr array_18 19 21 A_17_P17242359 - -
          Agilent Whole Chr array_18 19 21 A_17_P11413041 - -
          Agilent Whole Chr array_18 19 21 A_17_P11413042 - -
          Agilent Whole Chr array_18 19 21 A_17_P32075511 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32075505 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11413045 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17242360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11413044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11413043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11413042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11413041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32075519 - -
          Agilent Whole Chr array_18 19 21 A_17_P32075511 - -
          Agilent Whole Chr array_18 19 21 A_17_P11413044 - -
          Agilent Whole Chr array_18 19 21 A_17_P11413045 - -
          Agilent Whole Chr array_18 19 21 A_17_P17242359 - -
          Agilent Whole Chr array_18 19 21 A_17_P32075505 - -
          Agilent Whole Chr array_18 19 21 A_17_P32075519 - -
          Agilent Whole Chr array_18 19 21 A_17_P11413041 - -
          Agilent Whole Chr array_18 19 21 A_17_P17242360 - -
          Agilent Whole Chr array_18 19 21 A_17_P11413043 - -
          Agilent Whole Chr array_18 19 21 A_17_P17242357 - -
          Agilent Whole Chr array_18 19 21 A_17_P11413042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11413044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32075519 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11413041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17242360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11413043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11413045 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32075505 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11413042 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413050 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413004 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413006 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413043 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413042 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413049 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413046 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413055 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413053 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413013 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413041 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413005 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413010 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11413002 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17242356 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_198698_at - -
          Affymetrix Human Gene 1.0 ST Array 8070790 - -
          Affymetrix Human Gene 1.1 ST Array 8070789 - -
          Affymetrix Human Genome U219 Array 11742177_at - -
          Affymetrix Human Genome U219 Array 11742178_x_at - -
          Affymetrix Human Genome U219 Array 11742178_PM_x_at - -
          Affymetrix Human Genome U219 Array 11742177_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR21006325.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21006325.hg.1 NM_198698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21006325.hg.1 TC21000532.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2100008379.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008379.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008379.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100008379.hg.1 NM_198698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100008379.hg.1 NM_198698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100008379.hg.1 NM_198698.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100156785.hg.1 TC2100008379.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100156785.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100156785.hg.1 NM_198698.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100156785.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100156785.hg.1 NM_198698.1 Refseq
          Affymetrix Human Clariom D Assay TC2100008379.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008379.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008379.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100008379.hg.1 NM_198698.1 Refseq
          Affymetrix Human Clariom D Assay TC2100008379.hg.1 NM_198698.1 Refseq
          Affymetrix Human Clariom D Assay TC2100008379.hg.1 NM_198698.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100156787.hg.1 TC2100008379.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100156787.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100156787.hg.1 NM_198698.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100156787.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100156787.hg.1 NM_198698.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100156786.hg.1 TC2100008379.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100156786.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100156786.hg.1 NM_198698.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100156786.hg.1 ENST00000391618.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100156786.hg.1 NM_198698.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018980 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018980 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 709647 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1780422 NM_198698.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1780422 AB076360.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1780422 BC125198.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1780422 NM_198698.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1780422 AB076360.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1780422 BC125198.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1780422 NM_198698.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1780422 AB076360.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1780422 BC125198.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1780422 NM_198698.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1780422 AB076360.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1780422 BC125198.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1780422 NM_198698.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1780422 AB076360.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1780422 BC125198.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_38371743-S NM_198698.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_38371743-S AB076360.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_38371743-S BC125198.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_3119-S - -
          Illumina human-6 v1.0 expression beadchip 0003850017 - -
          Illumina humanRef-8 v1.0 expression beadchip 0003850017 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1780422 NM_198698.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1780422 AB076360.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1780422 BC125198.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1780422 NM_198698.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1780422 AB076360.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1780422 BC125198.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 6960398 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_3119 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1780422 NM_198698.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1780422 AB076360.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1780422 BC125198.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1780422 NM_198698.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1780422 AB076360.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1780422 BC125198.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1780422 NM_198698.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1780422 AB076360.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1780422 BC125198.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1780422 NM_198698.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1780422 AB076360.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1780422 BC125198.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1780422 NM_198698.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1780422 AB076360.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1780422 BC125198.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline