Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01651930_g1
          See other HSALR1 GE Assays ›
          Gene Symbol:
          HSALR1
          Gene Name:
          HSP90AB1 associated lncRNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 88731180 - 88741425 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01651930_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HSALR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_103774.1 2 - 3 849 106
          GenBank mRNA AK127064.1 - 1 - 2 1601 106
          AK294743.1 - 2 - 3 849 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600008838.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179493 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16928155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179486 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179503 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724849 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179491 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16928151 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16928161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179481 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179498 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16928159 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724885 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179499 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179485 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179505 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3419621 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 3057 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03197798 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03197794 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03197788 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7997831 - -
          Affymetrix Human Gene 1.0 ST Array 7997830 - -
          Affymetrix Human Gene 1.1 ST Array 7997829 - -
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 NR_103774.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 NR_103774.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 NR_103774.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008838.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165867.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165867.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165867.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165867.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165867.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165867.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165867.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165867.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165867.hg.1 TC1600008838.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008838.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165869.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165869.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165869.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165869.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165869.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165869.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165869.hg.1 TC1600008838.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600165865.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165865.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165865.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165865.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165865.hg.1 TC1600008838.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600165868.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165868.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165868.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165868.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165868.hg.1 ENST00000440406.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165868.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165868.hg.1 ENST00000567968.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165868.hg.1 NR_103774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165868.hg.1 TC1600008838.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline