| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01028501.hg.1 |
ENST00000360827.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01028501.hg.1 |
ENST00000445590.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01028501.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01028501.hg.1 |
TC01001831.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01028504.hg.1 |
ENST00000360827.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01028504.hg.1 |
ENST00000445590.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01028504.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01028504.hg.1 |
TC01001831.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000460763.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000460763.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
ENST00000460763.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000460763.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000460763.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
ENST00000460763.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011696.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000460763.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
TC0100011696.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100186172.hg.1 |
ENST00000460763.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
ENST00000360827.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
ENST00000391880.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
ENST00000434700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
ENST00000445590.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
ENST00000456298.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
NM_207468.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100186175.hg.1 |
TC0100011696.hg.1 |
Clariom D TC |