| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000415182.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000415182.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000415182.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000423581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000423581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000423581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000423581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000415182.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000415182.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000415182.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000423581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000423581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000423581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000423581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006765.hg.1 |
ENST00000453784.2 |
Ensembl |