| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005153.hg.1 |
ENST00000344526.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005153.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005153.hg.1 |
TC04000379.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005154.hg.1 |
ENST00000344526.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005154.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005154.hg.1 |
TC04000379.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005155.hg.1 |
ENST00000344526.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005155.hg.1 |
ENST00000502294.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005155.hg.1 |
ENST00000502441.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005155.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005155.hg.1 |
TC04000379.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400188385.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188385.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188385.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188385.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188385.hg.1 |
TC0400012807.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400188388.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188388.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188388.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188388.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188388.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188388.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188388.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188388.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188388.hg.1 |
TC0400012807.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400188386.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188386.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188386.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188386.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188386.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188386.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188386.hg.1 |
TC0400012807.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000344526.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000502294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000509633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
ENST00000512173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012807.hg.1 |
NM_214711.3 |
Refseq |