Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01667919_s1
          See other EMC3-AS1 GE Assays ›
          Gene Symbol:
          EMC3-AS1
          Gene Name:
          EMC3 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 9986893 - 10007004 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01667919_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EMC3-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_103821.1 3 94
          GenBank mRNA AK092352.1 - 2 94
          AK125558.1 - 4 1489 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300006573.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P384698 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P384698 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P384698 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8077730 - -
          Affymetrix Human Gene 1.0 ST Array 8077729 - -
          Affymetrix Human Gene 1.1 ST Array 8077728 - -
          Affymetrix HT HG-U133+ PM Array Plate 237005_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0300006615.hg.1 ENST00000449657.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006615.hg.1 ENST00000449657.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006615.hg.1 ENST00000449657.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000430621.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000430621.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145529.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145529.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145529.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145529.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145529.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145529.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145529.hg.1 TC0300006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300145546.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145546.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145546.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145546.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145546.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145546.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145546.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145546.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145546.hg.1 TC0300006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300145530.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145530.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145530.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145530.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145530.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145530.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145530.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145530.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145530.hg.1 TC0300006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300145538.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145538.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145538.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145538.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145538.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145538.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145538.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145538.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145538.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145538.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145538.hg.1 TC0300006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300145552.hg.1 ENST00000430621.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145552.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145552.hg.1 ENST00000430621.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145552.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145552.hg.1 TC0300006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300145550.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145550.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145550.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145550.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145550.hg.1 TC0300006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300145551.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145551.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145551.hg.1 TC0300006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0300006615.hg.1 ENST00000449657.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006615.hg.1 ENST00000449657.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006615.hg.1 ENST00000449657.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000430621.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000430621.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000438698.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006573.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145545.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145545.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145545.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145545.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145545.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145545.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145545.hg.1 ENST00000366264.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145545.hg.1 ENST00000424904.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145545.hg.1 ENST00000440568.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145545.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145545.hg.1 TC0300006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300145548.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145548.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145548.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145548.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145548.hg.1 TC0300006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300145549.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145549.hg.1 ENST00000430621.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145549.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145549.hg.1 ENST00000326237.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145549.hg.1 ENST00000430621.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145549.hg.1 NR_103821.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300145549.hg.1 TC0300006573.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 237005_PM_at - -
          Human Genome U133 Plus 2.0 Array 237005_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 237005_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 237005_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 237005_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 237005_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 237005_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 237005_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 237005_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 237005_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 237005_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 237005_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 237005_at10 - -
          Human Genome U95 Set 81221_at - -
          Human Genome U95 Set 81221_PM_at - -
          Human Genome U133 Set 237005_at - -
          Human Genome U133 Set 237005_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229833 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3289346 BC022834.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3289346 AK092352.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3289346 AK125558.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline