Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01669067_s1
          See other KRTAP4-6 GE Assays ›
          Gene Symbol:
          KRTAP4-6
          Gene Name:
          keratin associated protein 4-6
          Gene Aliases:
          KAP4.15, KAP4.6, KRTAP4-15, KRTAP4.15
          Chromosome Location:
          Chr.17: 89227 - 90338 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01669067_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KRTAP4-6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030976.2 NP_112238.1 162
          GenBank mRNA AJ406938.1 - 162
          AJ406945.1 - 162

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Homo sapiens 1700

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893159 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893163 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10321422 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893165 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893154 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3256257 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P417100 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8015220 - -
          Affymetrix Human Gene 1.0 ST Array 8015219 - -
          Affymetrix Human Gene 1.0 ST Array 8015231 - -
          Affymetrix Human Gene 1.1 ST Array 8015230 - -
          Affymetrix Human Gene 1.1 ST Array 8015218 - -
          Affymetrix HT HG-U133+ PM Array Plate 234683_PM_at - -
          Human Genome U133 Plus 2.0 Array 234683_PM_at - -
          Human Genome U133 Plus 2.0 Array 234683_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234683_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234683_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234683_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234683_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234683_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234683_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234683_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234683_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234683_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234683_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234683_at4 - -
          Human Genome U133 Set 234683_at - -
          Human Genome U133 Set 234683_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008317 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008317 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 235145 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline