Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01673896_s1
          See other KRTAP4-3 GE Assays ›
          Gene Symbol:
          KRTAP4-3
          Gene Name:
          keratin associated protein 4-3
          Gene Aliases:
          KAP4.3
          Chromosome Location:
          Chr.17: 41167231 - 41168172 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01673896_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KRTAP4-3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033187.1 NP_149443.1 1 693 146
          GenBank mRNA AJ406935.1 - 1 402 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700012410.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001494.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s)

          aging
          hair cycle

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16981597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893303 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10321548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893308 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10321535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10321539 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893319 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893316 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893310 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893317 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893301 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893306 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893293 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10321521 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10321527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10321550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10321519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893325 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893302 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893323 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30893314 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P346477 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3312743 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2969 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P346477 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2968 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 19085 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 3061 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 19571 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P346477 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3312743 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P346477 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3312743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893303 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321544 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893317 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321519 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893309 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321548 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16981597 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893314 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893301 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321543 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321550 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893310 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321521 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893313 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321539 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321545 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893316 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893307 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893302 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321539 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893301 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893313 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16981597 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321545 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893307 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321519 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321544 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893317 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893310 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893302 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893309 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893316 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321548 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321550 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893314 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30893303 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321521 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10321543 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P346477 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P346477 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 8463 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12649750 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321551 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321522 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321531 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321499 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16981599 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321527 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321496 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321518 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321491 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321493 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321524 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321508 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321537 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321494 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321540 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321504 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321506 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321529 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321520 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321534 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321503 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321547 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321501 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321550 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321549 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321536 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321487 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321542 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321544 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10321490 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_033187_at - -
          Affymetrix Human Gene 1.0 ST Array 8015241 - -
          Affymetrix Human Gene 1.1 ST Array 8015240 - -
          Affymetrix HT HG-U133+ PM Array Plate 234678_PM_at - -
          Affymetrix Human Genome U219 Array 11738795_at - -
          Affymetrix Human Genome U219 Array 11738795_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR17019739.hg.1 ENST00000391356.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019739.hg.1 NM_033187.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019739.hg.1 TC17001494.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700012410.hg.1 ENST00000391356.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012410.hg.1 ENST00000391356.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012410.hg.1 NM_033187.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012410.hg.1 NM_033187.1 Refseq
          Affymetrix Human Clariom D Assay TC1700012410.hg.1 ENST00000391356.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012410.hg.1 ENST00000391356.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012410.hg.1 NM_033187.1 Refseq
          Affymetrix Human Clariom D Assay TC1700012410.hg.1 NM_033187.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700206406.hg.1 ENST00000391356.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700206406.hg.1 NM_033187.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700206406.hg.1 ENST00000391356.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700206406.hg.1 NM_033187.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700206406.hg.1 TC1700012410.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700206407.hg.1 ENST00000391356.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700206407.hg.1 NM_033187.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700206407.hg.1 ENST00000391356.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700206407.hg.1 NM_033187.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700206407.hg.1 TC1700012410.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 234678_at - -
          Human Genome U133 Plus 2.0 Array 234678_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234678_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234678_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234678_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234678_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234678_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234678_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234678_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234678_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234678_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234678_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234678_at5 - -
          Human Genome U133 Set 234678_PM_at - -
          Human Genome U133 Set 234678_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008320 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008320 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1746434 NM_033187.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1746434 AJ406935.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1746434 NM_033187.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1746434 AJ406935.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1746434 NM_033187.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1746434 AJ406935.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1746434 NM_033187.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1746434 AJ406935.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1746434 NM_033187.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1746434 AJ406935.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1746434 NM_033187.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1746434 AJ406935.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1746434 NM_033187.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1746434 AJ406935.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline