Search
Search

KRTAP10-3| Gene Symbol: KRTAP10-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ566383.1 | - | 1 | 362 | 74 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | keratin associated protein 10-2 | KRTAP10-2 |
KAP10.2, KAP18-2, KAP18.2, KRTAP10.2, KRTAP18-2, KRTAP18.2 |
-6.3kb | Sense | Chr. 21 - 44550357 - 44551505 on NCBI Build GRCh38 |
| Downstream | keratin associated protein 10-4 | KRTAP10-4 |
KAP10.4, KAP18-4, KRTAP10.4, KRTAP18-4, KRTAP18.4 |
15.5kb | Anti-sense | Chr. 21 - 44573729 - 44575371 on NCBI Build GRCh38 |
Set Membership: |
|