Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01675489_s1
          See other OR2G2 GE Assays ›
          Gene Symbol:
          OR2G2
          Gene Name:
          olfactory receptor family 2 subfamily G member 2
          Gene Aliases:
          OR1-32
          Chromosome Location:
          Chr.1: 247588360 - 247589313 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01675489_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR2G2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001915.1 NP_001001915.1 1 903 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR2G2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003769 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003744 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003779 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003748 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003781 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003773 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003745 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003782 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929617 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003753 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003759 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003754 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929615 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003776 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21003747 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00929613 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3365077 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0170 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0171 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0172 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 37531 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P10592028 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3365077 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3365077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929598 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929604 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929601 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929592 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003770 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929591 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003752 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929592 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929601 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003770 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929591 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929604 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003752 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21003743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00929598 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P35543387 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929600 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929602 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929590 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929580 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929586 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929604 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929592 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929598 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929607 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929596 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929582 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929585 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929610 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00929588 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P35543387 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 156821 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 75014 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35543387 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929590 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929621 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929598 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929620 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929607 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929604 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929635 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929585 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929592 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929611 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929596 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929617 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929600 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929629 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929627 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929588 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929633 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929602 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929606 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929640 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929594 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929608 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929582 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929623 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929638 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00929612 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P35543387 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001001915_at - -
          Affymetrix Human Gene 1.0 ST Array 7911208 - -
          Affymetrix Human Gene 1.1 ST Array 7911207 - -
          Affymetrix Human Genome U219 Array 11742561_at - -
          Affymetrix Human Genome U219 Array 11742561_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A042303 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A042303 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 189869 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1772179 NM_001001915.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1772179 NM_001001915.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1772179 NM_001001915.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1772179 NM_001001915.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1772179 NM_001001915.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_12315-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1772179 NM_001001915.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1772179 NM_001001915.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 4390603 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_12315 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1772179 NM_001001915.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1772179 NM_001001915.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1772179 NM_001001915.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1772179 NM_001001915.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1772179 NM_001001915.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline