Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01869509_s1
          See other SPATA31D3 GE Assays ›
          Gene Symbol:
          SPATA31D3
          Gene Name:
          SPATA31 subfamily D member 3
          Gene Aliases:
          FAM75D3
          Chromosome Location:
          Chr.9: 81943500 - 81950094 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01869509_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SPATA31D3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207416.2 NP_997299.2 4 4129 128
          GenBank mRNA AK126070.1 - 1 2833 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900012145.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SPATA31D3

          Gene Ontology Categories:

          Function(s)

          spermatogenesis
          cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06774937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06774935 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06774938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16231942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27113796 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16231940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06774934 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 4094 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 189576 AK097698.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 189576 BC034552.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 189576 BC048325.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 189576 BC068579.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 189576 DQ991252.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 189576 NM_001085382.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 321644 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 112153 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 226156 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 37304 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3299696 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3577 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 59710 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02118955 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3299696 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3242609 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3299696 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774938 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774951 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774965 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774967 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774963 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774937 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774943 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774949 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774953 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774934 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774956 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774939 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774961 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774944 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774946 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774935 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06774970 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02118955 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8156101 - -
          Affymetrix Human Gene 1.0 ST Array 8156100 - -
          Affymetrix Human Gene 1.0 ST Array 8156099 - -
          Affymetrix Human Gene 1.0 ST Array 8156102 - -
          Affymetrix Human Gene 1.1 ST Array 8156098 - -
          Affymetrix Human Gene 1.1 ST Array 8156103 - -
          Affymetrix Human Genome U219 Array 11715328_PM_at - -
          Affymetrix Human Genome U219 Array 11715328_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 ENST00000603921.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 ENST00000603921.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 ENST00000603921.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 ENST00000603921.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 NM_001001670.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 NM_001001670.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 NM_001001670.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 NM_001001670.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 NM_001001670.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012146.hg.1 NM_001001670.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900184192.hg.1 NM_207416.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900184192.hg.1 NM_207416.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900184192.hg.1 TC0900012145.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 ENST00000344803.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 ENST00000603921.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 ENST00000603921.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 ENST00000603921.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 ENST00000603921.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 NM_001001670.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 NM_001001670.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 NM_001001670.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 NM_001001670.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 NM_001001670.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012146.hg.1 NM_001001670.2 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038838 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038838 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 218932 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1656779 AK126070.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1656779 NM_207416.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1656779 AK126070.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1656779 NM_207416.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline