Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01871537_s1
          See other SPRR2E GE Assays ›
          Gene Symbol:
          SPRR2E
          Gene Name:
          small proline rich protein 2E
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 153093135 - 153094526 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01871537_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SPRR2E
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024209.4 NP_001019380.2 2 608 127
          GenBank mRNA BC058018.1 - 2 625 127
          BG185829.1 - 1 84 127
          M20030.1 - 2 609 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100015851.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003255.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SPRR2E

          Gene Ontology Categories:

          Function(s) Process(es)

          epidermis development
          peptide cross-linking
          keratinization
          structural molecule activity
          protein binding
          structural constituent of epidermis

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571981 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15129683 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00514762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571952 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15129693 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571969 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571976 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00514746 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15129694 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15129691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00514754 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00514753 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20571957 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15129684 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3325704 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 15501 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 7055 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00165928 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3325704 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3325704 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571968 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571969 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129693 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571981 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571978 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129683 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129683 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571968 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571969 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571978 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15129693 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00514762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20571981 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P35310653 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P148949 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00165928 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2657361 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2055903 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2055903 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2657361 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00514746 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00514750 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00514748 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00514760 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00514753 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00514755 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15129656 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00514758 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00514754 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15129678 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00514762 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001024209_at - -
          Affymetrix Human Gene 1.0 ST Array 7920215 - -
          Affymetrix Human Gene 1.0 ST Array 7920216 - -
          Affymetrix Human Gene 1.1 ST Array 7920214 - -
          Affymetrix Human Genome U219 Array 11735693_x_at - -
          Affymetrix Human Genome U219 Array 11735693_PM_x_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0100015851.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015851.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015851.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015851.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015851.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015851.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015851.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015851.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015851.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01050155.hg.1 ENST00000368750.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050155.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050155.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01050155.hg.1 TC01003255.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01050156.hg.1 ENST00000368750.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050156.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050156.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01050156.hg.1 TC01003255.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01050158.hg.1 ENST00000368750.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050158.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01050158.hg.1 TC01003255.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100219043.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219043.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219043.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219043.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219043.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219043.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219043.hg.1 TC0100015851.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100219045.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219045.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219045.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219045.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219045.hg.1 TC0100015851.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100015851.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015851.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015851.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100015851.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015851.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015851.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015851.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015851.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015851.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219042.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219042.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219042.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219042.hg.1 ENST00000368750.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219042.hg.1 ENST00000368751.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219042.hg.1 NM_001024209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219042.hg.1 TC0100015851.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052250 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052250 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 105172 - -
          ABI Human Genome Survey Microarray Version 2.0 163047 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2211018 NM_001024209.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2211018 BC058018.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2211018 M20030.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1701239 M20030.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1701239 M20030.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2211018 NM_001024209.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2211018 BC058018.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2211018 M20030.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1701239 M20030.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2211018 NM_001024209.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2211018 BC058018.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2211018 M20030.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2211018 NM_001024209.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2211018 BC058018.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2211018 M20030.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_136986-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1701239 M20030.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1701239 M20030.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 730050 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_136986 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2211018 NM_001024209.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2211018 BC058018.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2211018 M20030.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2211018 NM_001024209.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2211018 BC058018.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2211018 M20030.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1701239 M20030.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1701239 M20030.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2211018 NM_001024209.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2211018 BC058018.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2211018 M20030.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1701239 M20030.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2211018 NM_001024209.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2211018 BC058018.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2211018 M20030.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline