Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01872667_s1
          See other SHANK2-AS3 GE Assays ›
          Gene Symbol:
          SHANK2-AS3
          Gene Name:
          SHANK2 antisense RNA 3
          Gene Aliases:
          C11orf76
          Chromosome Location:
          Chr.11: 70862790 - 70865115 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01872667_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SHANK2-AS3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_073536.1 3 1028 129
          GenBank mRNA BC004224.2 - 3 1028 3537945 129

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864351 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864342 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864335 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864373 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280630 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16469869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16469868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864379 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864381 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864376 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864383 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864348 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280646 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16469876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280633 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864371 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864334 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864357 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864340 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16469873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16469875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864396 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864363 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864330 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864390 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864368 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864374 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864392 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864380 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864386 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864364 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864332 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280632 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864395 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864329 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280639 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28280610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864370 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864359 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864353 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864341 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07864398 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 27265 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 59997 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3397161 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P244447 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7942231 - -
          Affymetrix Human Gene 1.0 ST Array 7942230 - -
          Affymetrix Human Gene 1.0 ST Array 7942229 - -
          Affymetrix Human Gene 1.1 ST Array 7942228 - -
          Affymetrix HT HG-U133+ PM Array Plate 224139_PM_at - -
          Human Genome U133 Plus 2.0 Array 224139_at - -
          Human Genome U133 Plus 2.0 Array 224139_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224139_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224139_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224139_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224139_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224139_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224139_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224139_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224139_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224139_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224139_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224139_at1 - -
          Human Genome U133 Set 224139_PM_at - -
          Human Genome U133 Set 224139_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 206102 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1669579 BC004224.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2168079 BC004224.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1669579 BC004224.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1669579 BC004224.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2168079 BC004224.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1669579 BC004224.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2168079 BC004224.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2168079 BC004224.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1669579 BC004224.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2168079 BC004224.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline