Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01899531_s1
          See other DEFB124 GE Assays ›
          Gene Symbol:
          DEFB124
          Gene Name:
          defensin beta 124
          Gene Aliases:
          DEFB-24
          Chromosome Location:
          Chr.20: 31465461 - 31475218 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01899531_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DEFB124
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037500.1 NP_001032589.1 2 152 82
          XM_017027782.1 XP_016883271.1 3 769 82
          GenBank mRNA AY122476.1 - 1 87 82
          DQ119827.1 - 2 152 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000009994.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000752.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DEFB124

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s)

          defense response to bacterium
          innate immune response

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777415 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130369 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130354 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777411 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130348 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777420 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130364 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777426 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130353 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130371 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777450 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130374 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130359 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130351 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777408 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130347 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130358 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130340 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130363 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17186371 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130367 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777434 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11130373 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31777432 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 768732 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 272410 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 1245 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 185250 NM_004928.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 185250 Y11392.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 185250 Z93322.1 genbank_mrna_human
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3316522 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1232 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1233 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 10302 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P34681095 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3316522 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3316522 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777444 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777442 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777450 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130372 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777445 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17186371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130366 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777449 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130369 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777449 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777445 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130366 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130369 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777444 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17186371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130372 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777442 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777450 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P11130384 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17186373 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03505450 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P22379887 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03505450 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130385 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130353 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130341 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130421 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130419 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130339 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130405 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130351 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130358 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130366 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130364 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130379 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130415 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130401 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130393 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130386 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130373 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130346 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17186371 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130388 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130355 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130376 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130349 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130384 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130371 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130417 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130356 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130399 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130409 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17186379 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130413 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130344 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130407 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130397 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130395 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130390 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130369 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130374 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130403 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130362 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130424 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130382 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03505450 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001037500_at - -
          Affymetrix Human Gene 1.0 ST Array 8065568 - -
          Affymetrix Human Gene 1.0 ST Array 8065567 - -
          Affymetrix Human Gene 1.1 ST Array 8065566 - -
          Affymetrix HT HG-U133+ PM Array Plate 1568375_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1568377_PM_x_at - -
          Affymetrix Human Genome U219 Array 11738794_PM_x_at - -
          Affymetrix Human Genome U219 Array 11738794_x_at - -
          Affymetrix Human Genome U219 Array 11738793_at - -
          Affymetrix Human Genome U219 Array 11738793_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR20010063.hg.1 ENST00000317676.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010063.hg.1 NM_001037500.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20010063.hg.1 TC20000752.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20010064.hg.1 ENST00000317676.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010064.hg.1 NM_001037500.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20010064.hg.1 TC20000752.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2000009994.hg.1 ENST00000317676.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009994.hg.1 ENST00000317676.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009994.hg.1 NM_001037500.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009994.hg.1 NM_001037500.1 Refseq
          Affymetrix Human Clariom D Assay TC2000009994.hg.1 ENST00000317676.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009994.hg.1 ENST00000317676.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009994.hg.1 NM_001037500.1 Refseq
          Affymetrix Human Clariom D Assay TC2000009994.hg.1 NM_001037500.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171313.hg.1 ENST00000317676.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171313.hg.1 NM_001037500.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171313.hg.1 TC2000009994.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000171313.hg.1 ENST00000317676.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171313.hg.1 NM_001037500.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171315.hg.1 ENST00000317676.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171315.hg.1 NM_001037500.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000171315.hg.1 TC2000009994.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000171315.hg.1 ENST00000317676.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000171315.hg.1 NM_001037500.1 Refseq
          Human Genome U133 Plus 2.0 Array 1568375_PM_at - -
          Human Genome U133 Plus 2.0 Array 1568377_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 1568377_x_at - -
          Human Genome U133 Plus 2.0 Array 1568375_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_001037500_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568375_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568375_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568377_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568375_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568377_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568375_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568377_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568375_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568377_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568377_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568377_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568375_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568375_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568375_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568377_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568377_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568375_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568377_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568377_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568375_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568377_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1568375_at5 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A017536 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A017536 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 545905 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2104732 NM_001037500.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2104732 AY122476.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2104732 DQ119827.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2104732 NM_001037500.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2104732 AY122476.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2104732 DQ119827.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2104732 NM_001037500.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2104732 AY122476.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2104732 DQ119827.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2104732 NM_001037500.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2104732 AY122476.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2104732 DQ119827.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2104732 NM_001037500.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2104732 AY122476.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2104732 DQ119827.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2104732 NM_001037500.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2104732 AY122476.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2104732 DQ119827.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2104732 NM_001037500.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2104732 AY122476.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2104732 DQ119827.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2104732 NM_001037500.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2104732 AY122476.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2104732 DQ119827.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline