Search
Search

OSTCP1| Gene Symbol: OSTCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028496.1 | 3 | 864 | 156 | ||
| GenBank mRNA | BC024224.1 | - | 3 | 868 | 156 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P317450 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 39508 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 39661 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24685 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P202347 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P136231 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P317450 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P317450 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205373 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409349 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409378 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205348 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409344 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409357 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409366 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205353 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205352 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205381 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409384 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205384 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15882021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205383 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205398 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205392 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409381 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409358 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409387 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205374 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205392 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409378 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205373 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409366 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409381 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205381 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409358 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205352 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409384 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205348 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205384 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205398 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15882021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409357 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205374 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409387 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205353 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409344 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05205383 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25409349 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P304395 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P317450 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P136231 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P202347 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8130523 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8130526 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8130527 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8130524 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8130525 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8130522 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1553931_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11738571_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11738571_PM_x_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028460.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028460.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028460.hg.1 | TC06002267.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013734.hg.1 | ENST00000455507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013734.hg.1 | ENST00000455507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013734.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013734.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013734.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013734.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013734.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013734.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013734.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013734.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028459.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028459.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028459.hg.1 | TC06002267.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028461.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028461.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028461.hg.1 | TC06002267.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0600013734.hg.1 | ENST00000455507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013734.hg.1 | ENST00000455507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013734.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013734.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013734.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013734.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013734.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013734.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013734.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013734.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194673.hg.1 | ENST00000455507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194673.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194673.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194673.hg.1 | ENST00000455507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194673.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194673.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194673.hg.1 | TC0600013734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600194675.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194675.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194675.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194675.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194675.hg.1 | TC0600013734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600194676.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194676.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194676.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194676.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194676.hg.1 | TC0600013734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600194672.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194672.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194672.hg.1 | ENST00000522287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194672.hg.1 | NR_028496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194672.hg.1 | TC0600013734.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1553931_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553931_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553931_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553931_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553931_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553931_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553931_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553931_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553931_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553931_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553931_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553931_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553931_at5 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030756 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030755 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030756 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030755 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 218648 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3246544 | NR_028496.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3246544 | BC024224.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1764650 | NR_028496.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1764650 | BC024224.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1764650 | NR_028496.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1764650 | BC024224.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3246544 | NR_028496.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3246544 | BC024224.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1764650 | NR_028496.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1764650 | BC024224.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1764650 | NR_028496.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1764650 | BC024224.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1764650 | NR_028496.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1764650 | BC024224.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764650 | NR_028496.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764650 | BC024224.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764650 | NR_028496.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764650 | BC024224.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1764650 | NR_028496.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1764650 | BC024224.1 | human_transcriptome |