| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031379.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031379.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031379.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031379.hg.1 |
TC01002075.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031380.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031380.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031380.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031380.hg.1 |
TC01002075.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031378.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031378.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031378.hg.1 |
TC01002075.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100191196.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191196.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191196.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191196.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191196.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191196.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191196.hg.1 |
TC0100012421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100191197.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191197.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191197.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191197.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191197.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191197.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191197.hg.1 |
TC0100012421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100191195.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191195.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191195.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191195.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191195.hg.1 |
TC0100012421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000427857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000432963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
ENST00000446136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012421.hg.1 |
NR_027055.1 |
Refseq |