| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
7942524 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7942522 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7942523 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7942521 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
7942520 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1554609_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012965.hg.1 |
ENST00000505454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012965.hg.1 |
ENST00000505454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012965.hg.1 |
ENST00000505454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012965.hg.1 |
ENST00000505454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000607662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000607662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000607662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000607691.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
ENST00000607691.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013070.hg.1 |
ENST00000612462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202052.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202052.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202052.hg.1 |
TC1100013070.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100202055.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202055.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202055.hg.1 |
TC1100013070.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000501071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000512406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000607662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000607662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000607662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000607691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
ENST00000607691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010026.hg.1 |
NR_039984.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013070.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013070.hg.1 |
ENST00000612462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202054.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202054.hg.1 |
ENST00000612462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202054.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202054.hg.1 |
ENST00000612462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202054.hg.1 |
TC1100013070.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100202053.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202053.hg.1 |
ENST00000526036.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202053.hg.1 |
TC1100013070.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400012965.hg.1 |
ENST00000505454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012965.hg.1 |
ENST00000505454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012965.hg.1 |
ENST00000505454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012965.hg.1 |
ENST00000505454.1 |
Ensembl |
| Human Genome U133 Plus 2.0 Array |
1554609_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1554609_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1554609_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1554609_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1554609_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1554609_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1554609_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1554609_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1554609_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1554609_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1554609_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1554609_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1554609_at10 |
- |
- |