Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01932048_s1
          See other LOC284912 GE Assays ›
          Gene Symbol:
          LOC284912
          Gene Name:
          uncharacterized LOC284912
          Gene Aliases:
          -
          Chromosome Location:
          Chr.22: 35627576 - 35637934 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01932048_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC284912
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC001801.1 - 2 638 3355513 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200008572.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17279524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11489869 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 104037 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 101064 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 884912 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 24065 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2683 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 102463 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03621469 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03621476 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41520433 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41520418 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03621473 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03621476 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P41520418 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 224117_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2200008572.hg.1 ENST00000472240.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008572.hg.1 ENST00000472240.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008572.hg.1 ENST00000472240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200161660.hg.1 TC2200008572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200161660.hg.1 ENST00000472240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200161660.hg.1 ENST00000472240.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008572.hg.1 ENST00000472240.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008572.hg.1 ENST00000472240.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008572.hg.1 ENST00000472240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200161659.hg.1 TC2200008572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200161659.hg.1 ENST00000472240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200161659.hg.1 ENST00000472240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200161662.hg.1 TC2200008572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200161662.hg.1 ENST00000472240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200161662.hg.1 ENST00000472240.1 Ensembl
          Human Genome U133 Plus 2.0 Array 224117_at - -
          Human Genome U133 Plus 2.0 Array 224117_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224117_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224117_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224117_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224117_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224117_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224117_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224117_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224117_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224117_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224117_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224117_at3 - -
          Human Genome U133 Set 224117_PM_at - -
          Human Genome U133 Set 224117_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 233541 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1804213 BC001801.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1804213 BC001801.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1804213 BC001801.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1804213 BC001801.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1804213 BC001801.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1804213 BC001801.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1804213 BC001801.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1804213 BC001801.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1804213 BC001801.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline