Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01939618_s1
          See other ZNF818P GE Assays ›
          Gene Symbol:
          ZNF818P
          Gene Name:
          zinc finger protein 818, pseudogene
          Gene Aliases:
          ZNF818
          Chromosome Location:
          Chr.19: 53212948 - 53216102 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01939618_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZNF818P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_073396.1 1 2077 148
          GenBank mRNA AK128250.1 - 1 2047 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900008739.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638032 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638003 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17158221 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997260 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638008 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638014 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17158232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637964 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17158220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637972 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17158238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17158240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17158219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17158236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638027 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17158235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638004 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637970 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638024 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17158222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637998 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17158234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17158244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637995 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31638020 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10997255 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3217594 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 11977 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 15582 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 9329 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P234691 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P804267 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P804267 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P234691 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 174624 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41225898 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P34606329 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8030971 - -
          Affymetrix Human Gene 1.0 ST Array 8030970 - -
          Affymetrix Human Gene 1.1 ST Array 8030969 - -
          Affymetrix HT HG-U133+ PM Array Plate 238149_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 238148_PM_s_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000598377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000601072.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000601072.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 ENST00000601072.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 NR_073396.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 NR_073396.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 NR_073396.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 NR_073396.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 NR_073396.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008739.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168270.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168270.hg.1 TC1900008739.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900168270.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168271.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168271.hg.1 TC1900008739.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900168271.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168272.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168272.hg.1 TC1900008739.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900168272.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168268.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168268.hg.1 TC1900008739.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900168268.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168269.hg.1 ENST00000598377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900168269.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168269.hg.1 TC1900008739.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900168269.hg.1 ENST00000598377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900168269.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168273.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168273.hg.1 TC1900008739.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900168273.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000597550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000598377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000601072.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000601072.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 ENST00000601072.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 NR_073396.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008739.hg.1 NR_073396.1 Refseq
          Human Genome U133 Plus 2.0 Array 238148_s_at - -
          Human Genome U133 Plus 2.0 Array 238148_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 238149_PM_at - -
          Human Genome U133 Plus 2.0 Array 238149_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238148_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238148_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238148_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238148_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238148_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238149_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238149_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238149_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238149_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238148_s_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238148_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238148_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238149_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238149_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238149_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238149_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238148_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238149_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238149_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238149_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238148_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238148_s_at3 - -
          Human Genome U95 Set 62751_PM_at - -
          Human Genome U95 Set 62751_at - -
          Human Genome U133 Set 238149_at - -
          Human Genome U133 Set 238149_PM_at - -
          Human Genome U133 Set 238148_PM_s_at - -
          Human Genome U133 Set 238148_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1654858 AK128250.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1654858 AK128250.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1654858 AK128250.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1654858 AK128250.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1654858 AK128250.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1654858 AK128250.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1654858 AK128250.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1654858 AK128250.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline