| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8070327 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8070325 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8070326 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8070322 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8070323 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8070324 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8070321 |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000615324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000615324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000626259.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000626259.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000626259.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
ENST00000626259.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
TC2100008155.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154970.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000440379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000613045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000615324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000615324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000623098.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000626259.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000626259.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000626259.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
ENST00000626259.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008155.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154949.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154949.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154949.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154949.hg.1 |
ENST00000626259.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154949.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154949.hg.1 |
TC2100008155.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100154949.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154949.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154949.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154949.hg.1 |
ENST00000626259.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154949.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154942.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154942.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154942.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154942.hg.1 |
TC2100008155.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100154942.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154942.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154942.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154943.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154943.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154943.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154943.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154943.hg.1 |
TC2100008155.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100154943.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154943.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154943.hg.1 |
ENST00000415824.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154943.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154941.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154941.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154941.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154941.hg.1 |
TC2100008155.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100154941.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154941.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154941.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154974.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154974.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154974.hg.1 |
NR_120405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100154974.hg.1 |
TC2100008155.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100154974.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154974.hg.1 |
ENST00000380931.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100154974.hg.1 |
NR_120405.1 |
Refseq |