Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01940019_s1
          See other ACOXL-AS1 GE Assays ›
          Gene Symbol:
          ACOXL-AS1
          Gene Name:
          ACOXL antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 111098341 - 111115588 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01940019_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ACOXL-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_122074.1 4 2706 94
          GenBank mRNA AK125994.1 - 3 2524 94
          OA987259.1 - 4 2706 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200013884.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01402566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21511966 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21511971 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01402575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01402574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21511979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21511969 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01402576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21511974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21511972 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01402572 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3359858 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 8899 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3359858 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3359858 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15782649 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00444316 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35811152 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15782659 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00444308 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35811169 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13351654 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00444312 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8054608 - -
          Affymetrix Human Gene 1.0 ST Array 8054609 - -
          Affymetrix Human Gene 1.0 ST Array 8054610 - -
          Affymetrix Human Gene 1.1 ST Array 8054607 - -
          Affymetrix Human Clariom D Assay PSR0200201677.hg.1 ENST00000376593.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201677.hg.1 NR_122074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201677.hg.1 ENST00000376593.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201677.hg.1 NR_122074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201677.hg.1 TC0200013884.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200201679.hg.1 ENST00000376593.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201679.hg.1 ENST00000418615.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201679.hg.1 NR_122074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201679.hg.1 ENST00000376593.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201679.hg.1 ENST00000418615.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201679.hg.1 NR_122074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201679.hg.1 TC0200013884.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200201680.hg.1 ENST00000418615.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201680.hg.1 NR_122074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201680.hg.1 ENST00000418615.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201680.hg.1 NR_122074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201680.hg.1 TC0200013884.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200201676.hg.1 ENST00000376593.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201676.hg.1 NR_122074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201676.hg.1 ENST00000376593.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201676.hg.1 NR_122074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201676.hg.1 TC0200013884.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200201681.hg.1 ENST00000418615.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201681.hg.1 NR_122074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201681.hg.1 ENST00000418615.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201681.hg.1 NR_122074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201681.hg.1 TC0200013884.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1709900 AK125994.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1709900 AK125994.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1709900 AK125994.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1709900 AK125994.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1709900 AK125994.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1709900 AK125994.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline