Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01940426_s1
          See other OR10G9 GE Assays ›
          Gene Symbol:
          OR10G9
          Gene Name:
          olfactory receptor family 10 subfamily G member 9
          Gene Aliases:
          OR10G10P
          Chromosome Location:
          Chr.11: 124023013 - 124023948 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01940426_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR10G9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001953.1 NP_001001953.1 1 735 151

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100009369.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001130.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089503 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089515 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089506 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16512058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089497 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28517180 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3290792 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3336627 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 51154 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 5245 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P33071603 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3290792 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3336627 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3290792 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3336627 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089515 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089491 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089516 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089497 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089508 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089519 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089501 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517195 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517176 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517179 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089502 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089490 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089518 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517197 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089502 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089518 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517197 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089501 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089490 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517179 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089508 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089516 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089491 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089519 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517176 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089497 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28517195 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089515 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089489 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089491 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089547 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089487 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089499 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089503 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089533 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089540 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089476 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089505 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089548 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089531 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089537 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089542 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089530 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089479 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089518 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089484 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089511 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089482 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089544 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089474 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089496 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089494 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089491 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089528 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089481 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089534 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089472 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089488 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089516 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089490 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089507 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089543 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089519 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089500 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001001953_at - -
          Affymetrix Human Gene 1.0 ST Array 7944800 - -
          Affymetrix Human Gene 1.1 ST Array 7944799 - -
          Affymetrix Human Genome U219 Array 11742608_PM_x_at - -
          Affymetrix Human Genome U219 Array 11742608_x_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11014308.hg.1 ENST00000375024.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11014308.hg.1 NM_001001953.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11014308.hg.1 TC11001130.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100009369.hg.1 ENST00000375024.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009369.hg.1 NM_001001953.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012651.hg.1 ENST00000330487.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012651.hg.1 NM_001004463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009368.hg.1 ENST00000320891.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009368.hg.1 NM_001004462.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012651.hg.1 ENST00000330487.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012651.hg.1 NM_001004463.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100170228.hg.1 ENST00000375024.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100170228.hg.1 NM_001001953.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100170228.hg.1 ENST00000375024.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100170228.hg.1 NM_001001953.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100170228.hg.1 TC1100009369.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100009368.hg.1 ENST00000320891.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009368.hg.1 NM_001004462.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009369.hg.1 ENST00000375024.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009369.hg.1 NM_001001953.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047071 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047071 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1732251 NM_001001953.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1732251 NM_001001953.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1732251 NM_001001953.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1732251 NM_001001953.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1732251 NM_001001953.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_13582-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1732251 NM_001001953.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1732251 NM_001001953.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 7650347 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_13582 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1732251 NM_001001953.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1732251 NM_001001953.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1732251 NM_001001953.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1732251 NM_001001953.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1732251 NM_001001953.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline