Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01943480_g1
          See other OR5M1 GE Assays ›
          Gene Symbol:
          OR5M1
          Gene Name:
          olfactory receptor family 5 subfamily M member 1
          Gene Aliases:
          OR11-208, OST050
          Chromosome Location:
          Chr.11: 56612555 - 56613502 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01943480_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR5M1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004740.1 NP_001004740.1 1 887 136

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR5M1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219726 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219715 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219729 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219685 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810133 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219706 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219724 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219730 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810125 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810117 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219732 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219697 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219703 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219705 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219699 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219727 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810104 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07810149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219688 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3343496 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0471 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0472 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 9768 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3343496 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3343496 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219705 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219710 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219697 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219729 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219713 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219724 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219688 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219690 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219723 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219702 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219704 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219699 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219713 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219690 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219697 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219710 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219699 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219688 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219723 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219705 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219729 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07810122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219704 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219702 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219724 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810105 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810099 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810121 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810133 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810108 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810117 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810153 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810101 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810145 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810147 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810116 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810142 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810103 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810123 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810143 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810097 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810128 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810135 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810119 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810138 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810127 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810155 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810131 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810107 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810104 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810093 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810114 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810156 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810150 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810140 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810095 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07810111 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810155 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810153 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810114 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810137 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810119 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810121 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810169 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810133 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810163 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810111 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810127 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810142 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810147 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810131 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810105 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810117 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810140 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810160 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810150 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810107 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810135 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810167 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810123 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810108 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810116 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07810145 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001004740_at - -
          Affymetrix Human Gene 1.0 ST Array 7948154 - -
          Affymetrix Human Gene 1.0 ST Array 7948152 - -
          Affymetrix Human Gene 1.0 ST Array 7948153 - -
          Affymetrix Human Gene 1.1 ST Array 7948150 - -
          Affymetrix Human Genome U219 Array 11742568_at - -
          Affymetrix Human Genome U219 Array 11742568_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045175 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045175 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3240726 NM_001004740.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681709 - -
          HumanHT-12 V4.0 expression beadchip ILMN_3240726 NM_001004740.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3240726 NM_001004740.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3240726 NM_001004740.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_26791-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1681709 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1681709 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_26791 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_26791 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1681709 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline