Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01943937_s1
          See other OR5M11 GE Assays ›
          Gene Symbol:
          OR5M11
          Gene Name:
          olfactory receptor family 5 subfamily M member 11
          Gene Aliases:
          OR11-199
          Chromosome Location:
          Chr.11: 56542340 - 56543257 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01943937_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR5M11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005245.1 NP_001005245.1 1 663 163
          GenBank mRNA BC136951.1 - 1 687 9020943 163
          BC136953.1 - 1 687 9020945 163

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100010863.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001780.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR5M11

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219431 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219426 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219425 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219446 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219441 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16453523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219448 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16453524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219427 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219434 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219430 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809875 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3318272 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3283147 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0600 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0601 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 10158 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 37390 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3318272 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3283147 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3318272 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3283147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219439 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219448 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16453524 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219425 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809880 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219441 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219427 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809875 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809869 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809877 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219434 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219440 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219445 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219430 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219448 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16453524 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219425 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219430 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219427 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219440 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809877 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219439 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809880 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219434 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219445 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809869 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809875 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219441 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P02447937 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02447937 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 13009 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809879 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809867 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809877 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809870 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16453524 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809858 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809872 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809864 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809875 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809880 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809857 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809852 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809856 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809854 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809873 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809862 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809859 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001005245_at - -
          Affymetrix Human Gene 1.0 ST Array 7948146 - -
          Affymetrix Human Gene 1.0 ST Array 7948145 - -
          Affymetrix Human Gene 1.1 ST Array 7948144 - -
          Affymetrix Human Genome U219 Array 11735980_at - -
          Affymetrix Human Genome U219 Array 11735980_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11020686.hg.1 ENST00000528616.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11020686.hg.1 NM_001005245.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11020686.hg.1 TC11001780.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100010863.hg.1 ENST00000528616.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010863.hg.1 NM_001005245.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100182127.hg.1 ENST00000528616.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100182127.hg.1 NM_001005245.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100182127.hg.1 ENST00000528616.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100182127.hg.1 NM_001005245.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100182127.hg.1 TC1100010863.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100182126.hg.1 TC1100010863.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100010863.hg.1 ENST00000528616.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100010863.hg.1 NM_001005245.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045171 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045171 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1792254 NM_001005245.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1792254 BC136951.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1792254 BC136953.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1792254 NM_001005245.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1792254 BC136951.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1792254 BC136953.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1792254 NM_001005245.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1792254 BC136951.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1792254 BC136953.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1792254 NM_001005245.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1792254 BC136951.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1792254 BC136953.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1792254 NM_001005245.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1792254 BC136951.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1792254 BC136953.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_8324-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1792254 NM_001005245.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1792254 BC136951.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1792254 BC136953.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1792254 NM_001005245.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1792254 BC136951.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1792254 BC136953.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 7610746 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_8324 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1792254 NM_001005245.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1792254 BC136951.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1792254 BC136953.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1792254 NM_001005245.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1792254 BC136951.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1792254 BC136953.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1792254 NM_001005245.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1792254 BC136951.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1792254 BC136953.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1792254 NM_001005245.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1792254 BC136951.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1792254 BC136953.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1792254 NM_001005245.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1792254 BC136951.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1792254 BC136953.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline