| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001018.hg.1 |
ENST00000246115.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001018.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001018.hg.1 |
TC19000066.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001017.hg.1 |
ENST00000246115.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001017.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001017.hg.1 |
TC19000066.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
ENST00000591346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
ENST00000591346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146260.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146260.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146260.hg.1 |
TC1900006621.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900146262.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146262.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146262.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146262.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146262.hg.1 |
TC1900006621.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900146263.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146263.hg.1 |
ENST00000591346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146263.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146263.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146263.hg.1 |
ENST00000591346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146263.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146263.hg.1 |
TC1900006621.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
ENST00000591346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
ENST00000591346.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006621.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146261.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146261.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146261.hg.1 |
ENST00000246115.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900146261.hg.1 |
NM_003775.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900146261.hg.1 |
TC1900006621.hg.1 |
Clariom D TC |