Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02339060_s1
          See other OR5T3 GE Assays ›
          Gene Symbol:
          OR5T3
          Gene Name:
          olfactory receptor family 5 subfamily T member 3
          Gene Aliases:
          OR11-178, OR5T3Q
          Chromosome Location:
          Chr.11: 56252200 - 56253222 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02339060_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR5T3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004747.1 NP_001004747.1 1 943 124

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR5T3

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808617 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808620 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218106 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218051 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218093 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218061 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218053 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218067 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218099 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218078 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218094 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218076 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218064 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218077 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218092 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218047 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218101 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218105 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218080 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218102 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218103 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808624 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218079 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218107 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218098 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218108 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218073 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218075 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218068 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218090 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218089 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218066 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218069 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3213874 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3271226 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0482 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0483 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 4524 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 1737 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P32909829 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3213874 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3271226 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3213874 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3271226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218064 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218083 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808617 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808620 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218076 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808631 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218090 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218089 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218094 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808629 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218092 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218066 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808618 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218066 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218089 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808617 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218049 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808631 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808620 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808629 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218083 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218076 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218094 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808618 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218092 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218064 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218090 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808586 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808601 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808621 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808629 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808599 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808595 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808605 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808587 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808616 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808620 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808624 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808614 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808632 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808584 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808593 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808627 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808592 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808583 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808603 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808607 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808590 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808598 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808579 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808611 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808616 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808624 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808599 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808601 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16453204 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808605 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808619 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808633 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808629 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808638 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808592 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808586 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808642 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808627 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808587 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808620 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808611 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808595 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808583 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808617 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808603 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808635 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808640 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808590 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16453202 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808614 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16453206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808644 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808598 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808607 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001004747_at - -
          Affymetrix Human Gene 1.0 ST Array 7939974 - -
          Affymetrix Human Gene 1.1 ST Array 7939973 - -
          Affymetrix Human Genome U219 Array 11742522_PM_at - -
          Affymetrix Human Genome U219 Array 11742522_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046398 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046398 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1703749 NM_001004747.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1703749 NM_001004747.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1703749 NM_001004747.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1703749 NM_001004747.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1703749 NM_001004747.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_7260-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1703749 NM_001004747.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1703749 NM_001004747.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 1170014 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_7260 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1703749 NM_001004747.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1703749 NM_001004747.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1703749 NM_001004747.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1703749 NM_001004747.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1703749 NM_001004747.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline