Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02339221_g1
          See other OR8H3 GE Assays ›
          Gene Symbol:
          OR8H3
          Gene Name:
          olfactory receptor family 8 subfamily H member 3
          Gene Aliases:
          OR11-172
          Chromosome Location:
          Chr.11: 56122373 - 56123311 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02339221_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR8H3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005201.1 NP_001005201.1 1 645 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100007626.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000446.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR8H3

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16453096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217612 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217564 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16453093 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217608 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808133 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16453110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16453136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16453130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217619 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808137 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217620 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217557 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16453122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808134 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28217623 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3300858 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0567 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0568 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 4628 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P11099044 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3300858 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3300858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217561 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217602 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217570 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16453109 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217576 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217557 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217548 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217555 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808128 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217560 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217556 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808137 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217595 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217552 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217598 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16453093 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217600 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217564 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16453096 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217550 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217553 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217564 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16453109 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217602 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217555 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217550 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217548 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217552 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217560 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217595 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217600 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217570 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16453096 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217557 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217598 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16453093 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217553 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808128 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808137 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217576 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217556 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28217561 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808119 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808131 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808127 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808137 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808128 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808123 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808130 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808120 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808135 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808122 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808134 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808125 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808136 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808133 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808121 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808126 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16453143 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808138 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808131 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16453091 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808134 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808147 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808158 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808145 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808159 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808133 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808161 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16453144 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808155 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808140 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808152 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808164 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808150 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808121 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16453133 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808127 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808130 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808139 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808128 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808136 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16453136 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808157 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808143 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001005201_at - -
          Affymetrix Human Gene 1.0 ST Array 7939970 - -
          Affymetrix Human Gene 1.1 ST Array 7939969 - -
          Affymetrix Human Genome U219 Array 11742598_PM_x_at - -
          Affymetrix Human Genome U219 Array 11742598_x_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11005528.hg.1 ENST00000313472.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005528.hg.1 NM_001005201.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005528.hg.1 TC11000446.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100007625.hg.1 ENST00000313503.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007625.hg.1 ENST00000618136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007625.hg.1 ENST00000618136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007625.hg.1 ENST00000618136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007625.hg.1 NM_001005200.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100153820.hg.1 ENST00000313472.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153820.hg.1 NM_001005201.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100153820.hg.1 ENST00000313472.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153820.hg.1 NM_001005201.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100153820.hg.1 TC1100007626.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100007625.hg.1 ENST00000313503.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007625.hg.1 ENST00000618136.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007625.hg.1 ENST00000618136.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007625.hg.1 ENST00000618136.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007625.hg.1 NM_001005200.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046396 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046396 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1768161 NM_001005201.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1768161 NM_001005201.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1768161 NM_001005201.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1768161 NM_001005201.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1768161 NM_001005201.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_6708-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1768161 NM_001005201.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1768161 NM_001005201.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 10167 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_6708 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1768161 NM_001005201.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1768161 NM_001005201.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1768161 NM_001005201.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1768161 NM_001005201.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1768161 NM_001005201.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline