Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02339520_g1
          See other EIF2S2 GE Assays ›
          Gene Symbol:
          EIF2S2
          Gene Name:
          eukaryotic translation initiation factor 2 subunit beta
          Gene Aliases:
          EIF2, EIF2B, EIF2beta, PPP1R67, eIF-2-beta
          Chromosome Location:
          Chr.20: 34088309 - 34112243 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02339520_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EIF2S2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001316363.2 NP_001303292.1 4 - 5 556 147
          NM_001316364.2 NP_001303293.1 4 - 5 565 147
          NM_003908.5 NP_003899.2 4 - 5 565 147
          XM_017028118.2 XP_016883607.1 4 - 5 981 147
          GenBank mRNA AK313025.1 - 4 - 5 549 147
          BC000461.2 - 4 - 5 568 147
          BC007888.2 - 3 - 4 359 147
          BT009795.1 - 3 - 4 432 147
          BX418731.2 - 4 - 5 548 147
          BX429051.2 - 4 - 5 515 147
          CR456734.1 - 3 - 4 432 147
          M29536.1 - 4 - 5 548 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000008909.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000779.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene EIF2S2

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          formation of translation preinitiation complex
          male germ cell proliferation
          cytoplasmic translational initiation
          translation
          translational initiation
          male gonad development
          RNA binding
          mRNA binding
          translation initiation factor activity
          protein binding
          translation factor activity, RNA binding
          zinc ion binding
          translation initiation factor binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20010402.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010402.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20010402.hg.1 TC20000779.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000162089.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162089.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000162089.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162089.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000162089.hg.1 TC2000008909.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline