Search
Search

OR5M8| Gene Symbol: OR5M8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005282.1 | NP_001005282.1 | 1 | 850 | 122 | |
| GenBank mRNA | BC136978.1 | - | 1 | 875 | 9020970 | 122 |
| BC136980.1 | - | 1 | 875 | 9020972 | 122 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809667 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219207 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219197 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219219 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16453447 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809675 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16453457 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219211 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16453462 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809668 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219179 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809670 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16453445 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219205 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219170 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809664 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219209 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809660 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219195 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809652 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3343531 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0621 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0620 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 45567 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3343531 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3343531 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219193 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16453445 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16453447 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809661 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809667 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219188 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16453457 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219176 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219190 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219176 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219188 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809667 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809661 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219193 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16453457 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16453447 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16453445 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219190 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809670 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809657 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809652 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809658 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809654 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809665 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809667 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809655 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809660 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809669 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809656 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809668 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809670 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809661 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809650 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16453445 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809652 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809665 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809656 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809663 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809657 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809668 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809661 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809675 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809669 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809672 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16453446 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001005282_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7948138 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7948137 | - | - |
| Affymetrix Human Genome U219 Array | 11738736_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11738736_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010859.hg.1 | ENST00000327216.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010859.hg.1 | NM_001005282.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010861.hg.1 | ENST00000530227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010860.hg.1 | ENST00000526140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020682.hg.1 | ENST00000327216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020682.hg.1 | NM_001005282.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020682.hg.1 | TC11001779.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100010860.hg.1 | ENST00000526140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182119.hg.1 | TC1100010859.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100010859.hg.1 | ENST00000327216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010859.hg.1 | NM_001005282.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182120.hg.1 | ENST00000327216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182120.hg.1 | NM_001005282.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182120.hg.1 | ENST00000327216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182120.hg.1 | NM_001005282.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182120.hg.1 | TC1100010859.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100010861.hg.1 | ENST00000530227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182121.hg.1 | TC1100010859.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045169 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045169 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1803873 | BC136978.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1803873 | BC136980.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1803873 | NM_001005282.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2199971 | BC136978.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2199971 | BC136980.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2199971 | NM_001005282.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1803873 | BC136978.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1803873 | BC136980.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1803873 | NM_001005282.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1803873 | BC136978.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1803873 | BC136980.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1803873 | NM_001005282.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2199971 | BC136978.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2199971 | BC136980.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2199971 | NM_001005282.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1803873 | BC136978.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1803873 | BC136980.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1803873 | NM_001005282.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2199971 | BC136978.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2199971 | BC136980.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2199971 | NM_001005282.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2199971 | BC136978.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2199971 | BC136980.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2199971 | NM_001005282.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1803873 | BC136978.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1803873 | BC136980.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1803873 | NM_001005282.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_24192-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1803873 | BC136978.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1803873 | BC136980.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1803873 | NM_001005282.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1803873 | BC136978.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1803873 | BC136980.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1803873 | NM_001005282.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 240193 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_24192 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2199971 | BC136978.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2199971 | BC136980.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2199971 | NM_001005282.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2199971 | BC136978.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2199971 | BC136980.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2199971 | NM_001005282.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1803873 | BC136978.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1803873 | BC136980.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1803873 | NM_001005282.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1803873 | BC136978.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1803873 | BC136980.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1803873 | NM_001005282.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2199971 | BC136978.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2199971 | BC136980.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2199971 | NM_001005282.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1803873 | BC136978.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1803873 | BC136980.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1803873 | NM_001005282.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2199971 | BC136978.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2199971 | BC136980.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2199971 | NM_001005282.1 | human_transcriptome |