| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011104.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011104.hg.1 |
ENST00000538474.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011104.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011104.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011104.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011104.hg.1 |
TC09002888.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012200.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900185532.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185532.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185532.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900185532.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900185532.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900185532.hg.1 |
ENST00000361134.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185532.hg.1 |
ENST00000538474.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185532.hg.1 |
NM_001177316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900185532.hg.1 |
NM_001177317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900185532.hg.1 |
NM_080877.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900185532.hg.1 |
TC0900012200.hg.1 |
Clariom D TC |