Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02341807_s1
          See other FLJ41170 GE Assays ›
          Gene Symbol:
          FLJ41170
          Gene Name:
          uncharacterized LOC440200
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02341807_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLJ41170
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK123165.1 - 1 1334 121

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979921 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979892 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979894 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517613 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979909 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979905 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979910 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16734948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16734938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517593 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517619 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979903 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979918 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979895 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979902 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979900 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517617 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979906 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979897 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979901 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517624 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979957 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09517611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979952 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29979934 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 635030 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 82137 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02985227 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02985227 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XR_041855_at - -
          Affymetrix Human Gene 1.0 ST Array XR_041856_at - -
          Affymetrix Human Gene 1.0 ST Array XR_041854_at - -
          Affymetrix Human Gene 1.0 ST Array 7976829 - -
          Affymetrix Human Gene 1.1 ST Array 7976828 - -
          Affymetrix HT HG-U133+ PM Array Plate 232955_PM_at - -
          Human Genome U133 Plus 2.0 Array 232955_at - -
          Human Genome U133 Plus 2.0 Array 232955_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232955_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232955_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232955_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232955_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232955_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232955_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232955_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232955_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232955_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232955_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232955_at10 - -
          Human Genome U133 Set 232955_at - -
          Human Genome U133 Set 232955_PM_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1722632 AK123165.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1722632 AK123165.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1722632 AK123165.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1722632 AK123165.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1722632 AK123165.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1722632 AK123165.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1722632 AK123165.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1722632 AK123165.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1722632 AK123165.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline