| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8112567 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8124476 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112503 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8177545 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8124479 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112498 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112559 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8124481 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112472 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112473 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8124474 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8124477 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112475 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112499 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8124475 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8177551 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8177546 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112505 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112566 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8177670 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8177549 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8177673 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112568 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112496 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112497 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8177548 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8124472 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112470 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8124473 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112504 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112516 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112569 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8177547 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8124478 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112474 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112518 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112471 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8177550 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8105996 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8177669 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8124469 |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019958.hg.1 |
NR_024054.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019958.hg.1 |
NR_029426.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019958.hg.1 |
TC05003447.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500193290.hg.1 |
NR_029426.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500193290.hg.1 |
NR_029426.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500193290.hg.1 |
TC0500013184.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
215599_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
238446_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
206565_x_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
215599_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
215043_s_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
206565_PM_x_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
238446_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
215043_PM_s_at |
- |
- |
| Human Genome U133A 2.0 Array |
206565_PM_x_at |
- |
- |
| Human Genome U133A 2.0 Array |
215599_at |
- |
- |
| Human Genome U133A 2.0 Array |
215043_PM_s_at |
- |
- |
| Human Genome U133A 2.0 Array |
215043_s_at |
- |
- |
| Human Genome U133A 2.0 Array |
215599_PM_at |
- |
- |
| Human Genome U133A 2.0 Array |
206565_x_at |
- |
- |
| AffymetrixPathwork Diagnostics Pathchip |
pw546842 |
- |
- |
| Human Genome U95 Set |
32921_at |
- |
- |
| Human Genome U95 Set |
32921_PM_at |
- |
- |
| Human Genome U95 Set |
41643_PM_at |
- |
- |
| Human Genome U95 Set |
41643_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206565_x_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238446_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238446_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215599_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206565_x_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215599_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215599_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238446_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206565_x_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215599_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206565_x_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206565_x_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215599_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206565_x_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215599_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206565_x_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238446_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215599_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238446_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215599_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238446_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206565_x_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206565_x_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238446_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206565_x_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215599_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215599_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206565_x_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238446_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215599_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238446_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238446_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
238446_at10 |
- |
- |
| Human Genome U95 Set |
92162_r_at |
- |
- |
| Human Genome U95 Set |
92162_PM_r_at |
- |
- |
| Human Genome U95 Set |
77461_at |
- |
- |
| Human Genome U95 Set |
77461_PM_at |
- |
- |
| Human Genome U133 Set |
215599_at |
- |
- |
| Human Genome U133 Set |
215599_PM_at |
- |
- |
| Human Genome U133 Set |
215043_PM_s_at |
- |
- |
| Human Genome U133 Set |
215043_s_at |
- |
- |
| Human Genome U133 Set |
206565_PM_x_at |
- |
- |
| Human Genome U133 Set |
206565_x_at |
- |
- |
| Human Genome U133 Set |
238446_PM_at |
- |
- |
| Human Genome U133 Set |
238446_at |
- |
- |