Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02341971_s1
          See other OR10S1 GE Assays ›
          Gene Symbol:
          OR10S1
          Gene Name:
          olfactory receptor family 10 subfamily S member 1
          Gene Aliases:
          OR11-279
          Chromosome Location:
          Chr.11: 123976696 - 123977691 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02341971_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR10S1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004474.1 NP_001004474.1 1 845 88
          GenBank mRNA BC137037.1 - 1 935 9021033 88
          BC137038.1 - 1 935 9021034 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100012648.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11002402.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR10S1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089308 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089283 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089288 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516976 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516973 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516946 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089280 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516981 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516959 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516971 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089261 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516963 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089276 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516985 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516952 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516957 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089295 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08089285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516954 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28516982 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3381077 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0383 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0384 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0382 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 25785 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02538244 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3381077 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3381077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516982 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089283 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516977 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516971 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089285 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089299 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516960 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516963 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516959 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089274 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089288 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516973 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516977 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089283 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089274 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089288 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516960 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516971 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516982 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089285 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516973 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516963 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089299 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516959 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08089275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28516983 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089270 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089295 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089298 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089273 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089283 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089275 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089290 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089278 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089285 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089293 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089287 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08089274 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089285 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089265 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089304 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089293 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089251 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089257 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089275 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089302 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089272 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089256 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089290 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089295 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089278 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089298 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089249 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089267 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089300 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089306 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16512025 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089283 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089274 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089270 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089287 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089261 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089252 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08089259 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001004474_at - -
          Affymetrix Human Gene 1.0 ST Array 7952385 - -
          Affymetrix Human Gene 1.1 ST Array 7952384 - -
          Affymetrix Human Genome U219 Array 11738685_at - -
          Affymetrix Human Genome U219 Array 11738685_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1100012648.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012648.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012648.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012648.hg.1 NM_001004474.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11028803.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028803.hg.1 NM_001004474.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11028803.hg.1 TC11002402.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1100012648.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012648.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012648.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012648.hg.1 NM_001004474.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100197362.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100197362.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100197362.hg.1 TC1100012648.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100197363.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100197363.hg.1 NM_001004474.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100197363.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100197363.hg.1 NM_001004474.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100197363.hg.1 TC1100012648.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100197364.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100197364.hg.1 ENST00000531945.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100197364.hg.1 TC1100012648.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045819 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045819 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1768567 NM_001004474.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1768567 BC137037.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1768567 BC137038.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1768567 NM_001004474.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1768567 BC137037.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1768567 BC137038.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1768567 NM_001004474.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1768567 BC137037.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1768567 BC137038.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1768567 NM_001004474.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1768567 BC137037.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1768567 BC137038.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1768567 NM_001004474.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1768567 BC137037.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1768567 BC137038.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_21810-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1768567 NM_001004474.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1768567 BC137037.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1768567 BC137038.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1768567 NM_001004474.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1768567 BC137037.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1768567 BC137038.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 6180735 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_21810 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1768567 NM_001004474.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1768567 BC137037.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1768567 BC137038.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1768567 NM_001004474.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1768567 BC137037.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1768567 BC137038.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1768567 NM_001004474.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1768567 BC137037.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1768567 BC137038.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1768567 NM_001004474.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1768567 BC137037.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1768567 BC137038.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1768567 NM_001004474.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1768567 BC137037.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1768567 BC137038.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline