Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02342067_s1
          See other OR4D11 GE Assays ›
          Gene Symbol:
          OR4D11
          Gene Name:
          olfactory receptor family 4 subfamily D member 11
          Gene Aliases:
          OR4D11P
          Chromosome Location:
          Chr.11: 59503576 - 59504511 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02342067_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR4D11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004706.1 NP_001004706.1 1 359 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100007739.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000490.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR4D11

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230758 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820277 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16455950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230773 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230774 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230790 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820258 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230756 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230796 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230786 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230757 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07820267 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230777 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28230781 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3389898 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0440 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0439 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 19629 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02451418 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3389898 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3389898 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820262 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16455950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230777 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820255 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230787 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230796 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230795 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230790 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820277 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820279 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230803 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820249 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820279 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820277 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230795 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16455950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230790 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820249 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230777 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230803 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820255 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230796 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230787 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28230767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07820262 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820279 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820270 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820277 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820251 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820256 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820247 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820268 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820273 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820244 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820261 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820264 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820278 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820274 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820269 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820266 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820249 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820258 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07820254 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02451419 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820305 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820258 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16455950 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820302 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820256 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820270 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820278 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820247 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820315 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820268 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820249 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820273 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820301 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820311 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820242 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820277 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820313 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820264 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820254 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820307 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820238 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820240 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820266 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820251 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820243 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820253 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820261 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820274 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07820279 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001004706_at - -
          Affymetrix Human Gene 1.0 ST Array 7940188 - -
          Affymetrix Human Gene 1.0 ST Array 7940184 - -
          Affymetrix Human Gene 1.0 ST Array 7940183 - -
          Affymetrix Human Gene 1.1 ST Array 7940187 - -
          Affymetrix Human Gene 1.1 ST Array 7940182 - -
          Affymetrix Human Genome U219 Array 11742609_at - -
          Affymetrix Human Genome U219 Array 11742609_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11005906.hg.1 ENST00000313253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005906.hg.1 NM_001004706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005906.hg.1 TC11000490.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100007739.hg.1 ENST00000313253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007739.hg.1 NM_001004706.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007739.hg.1 ENST00000313253.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100007739.hg.1 NM_001004706.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100154517.hg.1 ENST00000313253.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100154517.hg.1 NM_001004706.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100154517.hg.1 ENST00000313253.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100154517.hg.1 NM_001004706.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100154517.hg.1 TC1100007739.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046447 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046447 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1779078 NM_001004706.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1779078 NM_001004706.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1779078 NM_001004706.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1779078 NM_001004706.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1779078 NM_001004706.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_26062-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1779078 NM_001004706.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1779078 NM_001004706.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 2510037 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_26062 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1779078 NM_001004706.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1779078 NM_001004706.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1779078 NM_001004706.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1779078 NM_001004706.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1779078 NM_001004706.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline