Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02342156_s1
          See other OR5M9 GE Assays ›
          Gene Symbol:
          OR5M9
          Gene Name:
          olfactory receptor family 5 subfamily M member 9
          Gene Aliases:
          OR11-190
          Chromosome Location:
          Chr.11: 56462469 - 56463401 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02342156_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR5M9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004743.1 NP_001004743.1 1 867 160

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100010856.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001777.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR5M9

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219004 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218998 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809528 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809497 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809506 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809515 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07809505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218996 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219005 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219014 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219032 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28219013 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3343526 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0478 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0477 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 33060 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3343526 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3343526 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219024 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809489 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219033 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219013 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809515 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219008 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809514 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809497 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809496 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219005 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809528 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809505 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219031 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218998 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809529 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809496 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219005 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219008 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809505 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809514 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809529 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809528 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218998 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809497 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219024 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219013 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809489 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07809515 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219031 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28219033 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809517 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809496 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809529 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809521 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809531 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809493 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809534 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809536 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809526 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809528 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809518 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809491 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809515 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809477 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809504 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809489 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809499 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809506 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809532 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809501 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809487 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809509 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809479 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809483 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809513 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809486 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809524 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07809508 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809554 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809531 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809545 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809532 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809537 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809513 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809501 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809553 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809540 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809524 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809504 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809515 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809491 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809493 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809517 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809543 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809489 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809526 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809508 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809509 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809506 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809559 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809541 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809563 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809499 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809496 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809549 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809561 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809521 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07809534 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001004743_at - -
          Affymetrix Human Gene 1.0 ST Array 7948132 - -
          Affymetrix Human Gene 1.1 ST Array 7948131 - -
          Affymetrix Human Genome U219 Array 11742615_PM_at - -
          Affymetrix Human Genome U219 Array 11742615_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1100010856.hg.1 ENST00000279791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010856.hg.1 NM_001004743.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11020679.hg.1 ENST00000279791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11020679.hg.1 NM_001004743.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11020679.hg.1 TC11001777.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1100010856.hg.1 ENST00000279791.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010856.hg.1 NM_001004743.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100182114.hg.1 ENST00000279791.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100182114.hg.1 NM_001004743.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100182114.hg.1 ENST00000279791.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100182114.hg.1 NM_001004743.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100182114.hg.1 TC1100010856.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045167 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045167 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1761042 NM_001004743.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1772190 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1761042 NM_001004743.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1761042 NM_001004743.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1761042 NM_001004743.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_2095-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1772190 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1772190 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 4200379 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_2095 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1761042 NM_001004743.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1761042 NM_001004743.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1761042 NM_001004743.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1772190 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1761042 NM_001004743.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline