Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02342171_s1
          See other OR5T1 GE Assays ›
          Gene Symbol:
          OR5T1
          Gene Name:
          olfactory receptor family 5 subfamily T member 1
          Gene Aliases:
          OR11-179, OR5T1P
          Chromosome Location:
          Chr.11: 56275639 - 56276619 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02342171_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR5T1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004745.1 NP_001004745.1 1 54 101
          GenBank mRNA BC136992.1 - 1 140 9020984 101
          BC136993.1 - 1 140 9020985 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100007631.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000449.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR5T1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-cell signaling
          detection of chemical stimulus involved in sensory perception of smell
          adenylate cyclase-activating adrenergic receptor signaling pathway
          adrenergic receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218131 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808697 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218166 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218167 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218129 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218137 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218144 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28218151 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07808698 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3271221 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0481 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 47348 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P39363209 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3271221 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3271221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218154 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218130 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218139 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808699 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218142 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218153 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808697 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218140 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218170 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808672 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808699 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218140 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218130 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218154 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218142 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218153 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808697 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07808672 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218170 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218139 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28218143 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808699 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808681 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808684 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808697 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808680 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808667 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808687 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808692 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808700 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808670 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808702 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808677 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808686 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808695 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808674 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07808704 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P39363209 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39363209 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808702 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808700 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808674 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808704 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808708 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808686 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808677 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808684 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808692 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16453221 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808687 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808670 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808680 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808712 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16453214 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808697 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808706 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808666 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808688 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808694 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07808672 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P39363209 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001004745_at - -
          Affymetrix Human Gene 1.0 ST Array 7939976 - -
          Affymetrix Human Gene 1.1 ST Array 7939975 - -
          Affymetrix Human Genome U219 Array 11738689_x_at - -
          Affymetrix Human Genome U219 Array 11738688_PM_s_at - -
          Affymetrix Human Genome U219 Array 11738687_at - -
          Affymetrix Human Genome U219 Array 11738688_s_at - -
          Affymetrix Human Genome U219 Array 11738689_PM_x_at - -
          Affymetrix Human Genome U219 Array 11738687_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11005532.hg.1 ENST00000313033.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005532.hg.1 NM_001004745.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005532.hg.1 TC11000449.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100007631.hg.1 ENST00000313033.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007631.hg.1 NM_001004745.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100153829.hg.1 ENST00000313033.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153829.hg.1 NM_001004745.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100153829.hg.1 ENST00000313033.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100153829.hg.1 NM_001004745.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100153829.hg.1 TC1100007631.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100153828.hg.1 TC1100007631.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100153830.hg.1 TC1100007631.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100007631.hg.1 ENST00000313033.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100007631.hg.1 NM_001004745.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046399 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046399 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1766698 NM_001004745.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1766698 BC136992.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1766698 BC136993.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1766698 NM_001004745.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1766698 BC136992.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1766698 BC136993.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1766698 NM_001004745.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1766698 BC136992.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1766698 BC136993.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1766698 NM_001004745.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1766698 BC136992.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1766698 BC136993.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1766698 NM_001004745.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1766698 BC136992.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1766698 BC136993.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_5519-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1766698 NM_001004745.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1766698 BC136992.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1766698 BC136993.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1766698 NM_001004745.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1766698 BC136992.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1766698 BC136993.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_5519 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_5519 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1766698 NM_001004745.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1766698 BC136992.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1766698 BC136993.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1766698 NM_001004745.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1766698 BC136992.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1766698 BC136993.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1766698 NM_001004745.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1766698 BC136992.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1766698 BC136993.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1766698 NM_001004745.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1766698 BC136992.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1766698 BC136993.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1766698 NM_001004745.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1766698 BC136992.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1766698 BC136993.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline