Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02379661_g1
          See other MT2A GE Assays ›
          Gene Symbol:
          MT2A
          Gene Name:
          metallothionein 2A
          Gene Aliases:
          MT-2, MT-II, MT2
          Chromosome Location:
          Chr.16: 56608584 - 56609497 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02379661_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MT2A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005953.5 NP_005944.1 3 329 82
          GenBank mRNA BC007034.1 - 1 332 82
          BC070289.1 - 1 326 82
          V00594.1 - 3 314 82
          X97260.1 - 3 256 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600007957.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular copper ion homeostasis
          cellular zinc ion homeostasis
          detoxification of copper ion
          cellular response to interleukin-3
          cellular response to erythropoietin
          negative regulation of growth
          cellular response to cadmium ion
          cellular response to copper ion
          cellular response to zinc ion
          protein binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035303 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30575246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035306 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16899387 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30575213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035278 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035288 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30575251 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30575250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16899388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035301 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035283 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035295 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30575255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30575247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30575217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035298 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30575226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30575218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30575253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035279 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30575211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10035281 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 134574 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 60606 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P106844 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P106844 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P361896 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 19134 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 9570 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 19316 - -
          Agilent Human Genome CGH Microarray 44B A_14_P100385 - -
          Agilent Human Genome CGH Microarray 44A A_14_P100385 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 58570 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P12466425 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P20476974 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P106844 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P106844 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P303874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035283 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035284 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035276 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035295 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035279 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035273 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035285 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035292 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16899381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035273 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035284 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16899381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035285 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035279 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035292 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035283 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035276 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10035295 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30575211 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P106844 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P361896 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P100385 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035290 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035279 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035275 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035298 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035282 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035277 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035284 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035285 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035292 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035287 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035273 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035289 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035278 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035296 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035283 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035271 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10035294 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16899378 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16899381 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16899379 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16899377 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16899380 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10035290 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10035291 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P100385 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P100385 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P106844 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P361896 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P252413 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 44009 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 79566 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P100385 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035320 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035289 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035323 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16899381 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035319 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035278 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035325 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035311 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035310 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035275 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035273 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035313 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035279 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035284 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16899378 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035287 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035309 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035270 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035283 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035285 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035295 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035331 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035282 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035292 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035277 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10035307 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P100385 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_005953_at - -
          Affymetrix Human Gene 1.0 ST Array 8095378 - -
          Affymetrix Human Gene 1.0 ST Array 7995784 - -
          Affymetrix Human Gene 1.0 ST Array 7995785 - -
          Affymetrix Human Gene 1.0 ST Array 7995786 - -
          Affymetrix Human Gene 1.0 ST Array 8095363 - -
          Affymetrix Human Gene 1.0 ST Array 8095379 - -
          Affymetrix Human Gene 1.1 ST Array 8095376 - -
          Affymetrix Human Gene 1.1 ST Array 7995783 - -
          Affymetrix Human Gene 1.1 ST Array 8095362 - -
          Affymetrix HT HG-U133+ PM Array Plate 212185_PM_x_at - -
          Affymetrix Human Genome U219 Array 11753900_PM_x_at - -
          Affymetrix Human Genome U219 Array 11753900_x_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1800007593.hg.1 ENST00000583846.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007593.hg.1 ENST00000583846.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 ENST00000245185.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 ENST00000561491.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 ENST00000562017.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 ENST00000563985.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 ENST00000567300.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 NM_005953.4 Refseq
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 ENST00000245185.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 ENST00000561491.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 ENST00000562017.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 ENST00000563985.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 ENST00000567300.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 NM_005953.4 Refseq
          Affymetrix Human Clariom D Assay PSR1600158038.hg.1 TC1600007957.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 212185_x_at - -
          Human Genome U133 Plus 2.0 Array 212185_PM_x_at - -
          Human Genome U133A 2.0 Array 212185_PM_x_at - -
          Human Genome U133A 2.0 Array 212185_x_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_005953_at - -
          AffymetrixPathwork Diagnostics Pathchip pw537069 - -
          Human Genome U95 Set 39081_at - -
          Human Genome U95 Set 39081_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 212185_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 212185_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 212185_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 212185_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 212185_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 212185_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 212185_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 212185_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 212185_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 212185_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 212185_x_at7 - -
          Human Genome U95 Set 89884_at - -
          Human Genome U95 Set 89884_PM_at - -
          Human Genome U133 Set 212185_x_at - -
          Human Genome U133 Set 212185_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054615 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054615 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1686664 V00594.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1690957 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1686664 V00594.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1686664 V00594.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1686664 V00594.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31543214-S NM_005953.3 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31543214-S BC007034.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31543214-S BC070289.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31543214-S CR621046.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31543214-S CR625648.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_11198-S - -
          Illumina human-6 v1.0 expression beadchip 0007050592 - -
          Illumina humanRef-8 v1.0 expression beadchip 0007050592 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1690957 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1690957 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_11198 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_11198 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1686664 V00594.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1686664 V00594.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1686664 V00594.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1690957 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1686664 V00594.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline