| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362560 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362530 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362582 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940849 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940812 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362570 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940796 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940855 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362553 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940865 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362561 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940803 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362546 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940807 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362577 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940863 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362544 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940805 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940850 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362572 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940790 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362547 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940825 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940789 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362551 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940810 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940791 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362531 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362569 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362555 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940851 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940787 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940844 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362525 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362587 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940817 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362568 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940818 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362583 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362584 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362590 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362542 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940859 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940848 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940834 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362535 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362527 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362586 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940797 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16995911 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940806 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362564 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940795 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362562 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362565 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362573 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362566 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362576 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940821 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30940804 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362592 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362571 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16995912 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362541 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362556 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10362529 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
134367 |
NM_001099408.1 |
refseq_mrna_human_NM |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3543288 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
3696 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3543288 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3543288 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03257915 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P03257915 |
- |
- |