Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02384135_gH
          See other HSD3BP4 GE Assays ›
          Gene Symbol:
          HSD3BP4
          Gene Name:
          hydroxy-delta-5-steroid dehydrogenase, 3 beta, pseudogene 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 119563880 - 119572576 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02384135_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HSD3BP4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033781.1 1 7669 93
          GenBank mRNA AF252254.1 - 1 254 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100009617.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001041.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          dehydratase

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492159 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492180 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492185 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492163 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526434 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492171 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492144 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106395 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492166 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492135 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526454 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106384 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526460 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106379 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526544 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106380 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106477 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526441 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492187 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106511 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00492170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15106528 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20526446 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 140642 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 153654 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 968690 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 779584 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 190347 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 886911 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 385872 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 567532 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 568578 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 950484 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 219158 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 284706 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 326087 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 286348 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 336242 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 359913 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 64216 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 33065 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 187027 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 42956 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 54440 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 224562 AK124514.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 224562 BC063875.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 192254 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 277490 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 33568 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 344537 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 292968 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 296363 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 115832 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 141247 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 33389 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 284830 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 131621 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 218784 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 225634 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 4625 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 3453 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 13662 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 311036 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 72465 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 199168 AB039920.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 199168 AJ404617.2 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 199168 BC098159.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 199168 BC098256.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 199168 BC098319.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 199168 BC098344.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 13407 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 100483 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 148709 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 123117 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 299386 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 356101 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 299550 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 2892 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P350397 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 157352 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 126406 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 61924 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 76711 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10302004 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00158314 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15277557 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35289002 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P17810140 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00158316 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00158304 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15277572 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10303057 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00158307 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15277567 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10300023 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00158323 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10300970 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P17808146 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P17810132 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P10302004 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00158323 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00158307 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P10300970 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P15277572 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P15277557 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7904430 - -
          Affymetrix Human Gene 1.1 ST Array 7904429 - -
          Affymetrix Human Transcriptome Array 2.0 PSR01016496.hg.1 TC01001041.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01016496.hg.1 NR_033781.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01016497.hg.1 TC01001041.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01016497.hg.1 ENST00000335580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016497.hg.1 NR_033781.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01016498.hg.1 TC01001041.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01016498.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169229.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169229.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169229.hg.1 TC0100009617.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100169231.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169231.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169231.hg.1 TC0100009617.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100169233.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169233.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169233.hg.1 TC0100009617.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100169232.hg.1 ENST00000511509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100169232.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169232.hg.1 ENST00000511509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100169232.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169232.hg.1 TC0100009617.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100169230.hg.1 ENST00000511509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100169230.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169230.hg.1 ENST00000511509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100169230.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169230.hg.1 TC0100009617.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100169234.hg.1 ENST00000511509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100169234.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169234.hg.1 ENST00000511509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100169234.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169234.hg.1 TC0100009617.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100169235.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169235.hg.1 NR_033781.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100169235.hg.1 TC0100009617.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 215665_PM_at - -
          Human Genome U133 Plus 2.0 Array 215665_at - -
          Human Genome U133A 2.0 Array 215665_PM_at - -
          Human Genome U133A 2.0 Array 215665_at - -
          Human Genome U133 Set 215665_at - -
          Human Genome U133 Set 215665_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029106 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029106 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 195297 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1775655 NR_033781.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1775655 NR_033781.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1775655 NR_033781.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1775655 NR_033781.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline