| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019908.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019908.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019908.hg.1 |
TC06001672.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600179959.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179959.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179959.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179959.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179959.hg.1 |
TC0600011657.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
ENST00000437635.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011657.hg.1 |
NM_001010903.4 |
Refseq |