Search
Search

SPANXN2| Gene Symbol: SPANXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009615.2 | NP_001009615.1 | 2 | 214 | 163 | |
| GenBank mRNA | DQ336111.1 | - | 2 | 214 | 163 | |
| DQ336112.1 | - | 2 | 214 | 163 | ||
| DQ336113.1 | - | 2 | 214 | 163 | ||
| DQ336114.1 | - | 2 | 214 | 163 | ||
| DQ336115.1 | - | 2 | 214 | 163 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P12000887 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P12000882 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17406697 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32735334 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32735329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P12000891 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32735295 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32735300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32735302 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17406695 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32735297 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 147021 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 529575 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 908615 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 20617 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 268817 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 299204 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3422085 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0812 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0811 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 23777 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P03784864 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3422085 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3422085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32735334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17406697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32735329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32735331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32735335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12000892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17406695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12000892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32735334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17406697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32735329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32735331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32735335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17406695 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P12000892 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P12000891 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P12000890 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03784864 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03784864 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12000892 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12000886 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12000891 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12000881 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12000888 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12000884 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12000895 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12000893 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12000879 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P12000890 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03784864 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001009615_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8175580 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8175581 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8175579 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8175582 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8175578 | - | - |
| Affymetrix Human Genome U219 Array | 11740894_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11740894_x_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019102.hg.1 | TC0X001447.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019102.hg.1 | ENST00000370498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019102.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011014.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011014.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011014.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011014.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011014.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011014.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019100.hg.1 | TC0X001447.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019100.hg.1 | ENST00000370498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019100.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019101.hg.1 | TC0X001447.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019101.hg.1 | ENST00000370498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019101.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019103.hg.1 | TC0X001447.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019103.hg.1 | ENST00000370498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019103.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011014.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011014.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011014.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011014.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011014.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011014.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174759.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00174759.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174759.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00174759.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174759.hg.1 | TC0X00011014.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00174760.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00174760.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174760.hg.1 | ENST00000598475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00174760.hg.1 | NM_001009615.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174760.hg.1 | TC0X00011014.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040491 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040491 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1720259 | DQ336111.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1720259 | DQ336111.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1720259 | DQ336111.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1720259 | DQ336111.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1720259 | DQ336111.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_15295-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1720259 | DQ336111.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1720259 | DQ336111.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 7400113 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_15295 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1720259 | DQ336111.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1720259 | DQ336111.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1720259 | DQ336111.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1720259 | DQ336111.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1720259 | DQ336113.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1720259 | DQ336114.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1720259 | DQ336115.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1720259 | NM_001009615.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1720259 | DQ336111.1 | human_transcriptome |