| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135381 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135409 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697034 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135457 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697011 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696920 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697008 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696963 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697013 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697047 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696955 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135543 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697030 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696929 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135465 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696932 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696974 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697027 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697004 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697009 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16605911 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696910 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697052 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135494 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697003 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696956 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16605931 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135542 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697005 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697017 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697010 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696954 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697050 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135434 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135544 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696967 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697028 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696934 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135532 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135477 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696915 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697023 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135487 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696943 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696949 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696986 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696946 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135483 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135479 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696971 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696938 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135380 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697044 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697035 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696979 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696923 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697016 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696922 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697014 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696994 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696969 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696978 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696927 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135444 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697026 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697007 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135533 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696999 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697036 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135435 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697025 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696911 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696914 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696919 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696952 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135524 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135534 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697015 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696972 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697031 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696995 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697048 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696950 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135420 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696921 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135472 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696958 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696908 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697006 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696906 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697000 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696983 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697012 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135401 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697020 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135473 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696942 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696948 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696996 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696939 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696989 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696992 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135449 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135461 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16605940 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135513 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696917 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08696988 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29135423 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08697018 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
42403 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
481302 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
822559 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
970785 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
532709 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
77281 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
331682 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
208455 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
326792 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
380821 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
146259 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
161929 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
105813 |
BC000988.2 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
211018 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
61924 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
397306 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
335657 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
267143 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
124980 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
93472 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
209183 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
282928 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
169063 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
412489 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
366275 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
322317 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
162054 |
AB040879.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
162054 |
AL390162.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
162054 |
BC002607.2 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
162054 |
NM_020836.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
290879 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
198769 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
292246 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
1234 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P19729408 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3259077 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_16_P19729408 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P19729408 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P19729421 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
160686 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
134314 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
51790 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P39866094 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19729408 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P02729292 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19729396 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P19546301 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19729421 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P19729408 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P19729421 |
- |
- |